Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69109738-69109989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAS3
Entrez Gene ID
3038
Gene Name
hyaluronan synthase 3
Gene Aliases
-
UniGene
Hs.592069
Gene Chromosome Location
Chr.16: 69105564-69118719 on Build GRCh38
Transcripts
NM_001199280.1 NM_005329.2 NM_138612.2 XM_005255921.2 XM_011523061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972847 c.391T>C p.Y131H Chr.16: 69109786-69109786 on Build GRCh38
3272045 c.582C>T p.R194R Chr.16: 69109977-69109977 on Build GRCh38
6995170 c.497_500delGCCT p.L167fs*71 Chr.16: 69109892-69109895 on Build GRCh38
3272035 c.438C>T p.A146A Chr.16: 69109833-69109833 on Build GRCh38
972851 c.517C>T p.R173C Chr.16: 69109912-69109912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147358041 Chr.16: 69109947-69109947 on Build GRCh38
rs772651106 Chr.16: 69109827-69109827 on Build GRCh38
rs138020977 Chr.16: 69109854-69109854 on Build GRCh38
rs370216657 Chr.16: 69109818-69109818 on Build GRCh38
rs769971665 Chr.16: 69109771-69109771 on Build GRCh38
rs140402676 Chr.16: 69109845-69109845 on Build GRCh38
rs2232229 Chr.16: 69109913-69109913 on Build GRCh38
rs764759873 Chr.16: 69109823-69109823 on Build GRCh38
rs776448023 Chr.16: 69109877-69109877 on Build GRCh38
rs918756140 Chr.16: 69109907-69109907 on Build GRCh38
rs746052927 Chr.16: 69109834-69109834 on Build GRCh38
rs909522232 Chr.16: 69109916-69109916 on Build GRCh38
rs895626337 Chr.16: 69109820-69109820 on Build GRCh38
rs773716953 Chr.16: 69109833-69109833 on Build GRCh38
rs745597687 Chr.16: 69109962-69109962 on Build GRCh38
rs986765060 Chr.16: 69109857-69109857 on Build GRCh38
rs752026030 Chr.16: 69109830-69109830 on Build GRCh38
rs774986790 Chr.16: 69109887-69109887 on Build GRCh38
rs868753885 Chr.16: 69109795-69109795 on Build GRCh38
rs756415962 Chr.16: 69109921-69109921 on Build GRCh38
rs149097421 Chr.16: 69109883-69109883 on Build GRCh38
rs200060062 Chr.16: 69109816-69109816 on Build GRCh38
rs1056687956 Chr.16: 69109961-69109961 on Build GRCh38
rs148157588 Chr.16: 69109954-69109954 on Build GRCh38
rs766743263 Chr.16: 69109914-69109914 on Build GRCh38
rs750763427 Chr.16: 69109919-69109919 on Build GRCh38
rs879038572 Chr.16: 69109932-69109932 on Build GRCh38
rs746954845 Chr.16: 69109946-69109946 on Build GRCh38
rs770605226 Chr.16: 69109873-69109873 on Build GRCh38
rs770097327 Chr.16: 69109842-69109842 on Build GRCh38
rs749439912 Chr.16: 69109931-69109931 on Build GRCh38
rs777810606 Chr.16: 69109937-69109937 on Build GRCh38
rs145859833 Chr.16: 69109884-69109884 on Build GRCh38
rs774466945 Chr.16: 69109777-69109777 on Build GRCh38
rs767800507 Chr.16: 69109917-69109917 on Build GRCh38
rs764717797 Chr.16: 69109909-69109909 on Build GRCh38
rs577714323 Chr.16: 69109819-69109819 on Build GRCh38
rs752155032 Chr.16: 69109824-69109824 on Build GRCh38
rs773069546 Chr.16: 69109783-69109783 on Build GRCh38
rs566902020 Chr.16: 69109779-69109779 on Build GRCh38
rs766533894 Chr.16: 69109840-69109842 on Build GRCh38
rs751988494 Chr.16: 69109912-69109912 on Build GRCh38
rs142670939 Chr.16: 69109930-69109930 on Build GRCh38
rs762444635 Chr.16: 69109905-69109905 on Build GRCh38
rs1018555293 Chr.16: 69109872-69109872 on Build GRCh38
rs150384694 Chr.16: 69109850-69109850 on Build GRCh38
rs754993379 Chr.16: 69109935-69109935 on Build GRCh38
rs758922256 Chr.16: 69109825-69109825 on Build GRCh38
rs762433655 Chr.16: 69109806-69109806 on Build GRCh38
rs766115426 Chr.16: 69109811-69109811 on Build GRCh38
rs151215377 Chr.16: 69109782-69109782 on Build GRCh38
rs749320561 Chr.16: 69109849-69109849 on Build GRCh38

View Full Product Details