Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212064912-212065145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
UniGene
Hs.656473
Gene Chromosome Location
Chr.1: 212035553-212105006 on Build GRCh38
Transcripts
NM_001286229.1 NM_001286230.1 NM_016448.3 XM_011509614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349955 c.612G>A p.Q204Q Chr.1: 212065002-212065002 on Build GRCh38
4934711 c.527-2A>G p.? Chr.1: 212064915-212064915 on Build GRCh38
5845384 c.610C>T p.Q204* Chr.1: 212065000-212065000 on Build GRCh38
7089718 c.590C>A p.P197H Chr.1: 212064980-212064980 on Build GRCh38
4027917 c.561A>G p.G187G Chr.1: 212064951-212064951 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747196342 Chr.1: 212065058-212065059 on Build GRCh38
rs780968796 Chr.1: 212064959-212064959 on Build GRCh38
rs367567616 Chr.1: 212065072-212065072 on Build GRCh38
rs140634631 Chr.1: 212065004-212065004 on Build GRCh38
rs746406987 Chr.1: 212064991-212064993 on Build GRCh38
rs769115066 Chr.1: 212065043-212065043 on Build GRCh38
rs948702569 Chr.1: 212065038-212065038 on Build GRCh38
rs958234122 Chr.1: 212064977-212064977 on Build GRCh38
rs772090987 Chr.1: 212065060-212065060 on Build GRCh38
rs373660724 Chr.1: 212065071-212065071 on Build GRCh38
rs769005657 Chr.1: 212065061-212065079 on Build GRCh38
rs189644922 Chr.1: 212065019-212065019 on Build GRCh38
rs776967527 Chr.1: 212065079-212065079 on Build GRCh38
rs780303898 Chr.1: 212065054-212065055 on Build GRCh38
rs1044477142 Chr.1: 212065053-212065053 on Build GRCh38
rs773292910 Chr.1: 212065068-212065068 on Build GRCh38
rs923882764 Chr.1: 212064976-212064976 on Build GRCh38
rs765997063 Chr.1: 212064980-212064980 on Build GRCh38
rs758531289 Chr.1: 212065053-212065056 on Build GRCh38
rs748685719 Chr.1: 212065052-212065052 on Build GRCh38

View Full Product Details