Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133558074-133558301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYCE1
Entrez Gene ID
93426
Gene Name
synaptonemal complex central element protein 1
Gene Aliases
C10orf94, CT76, POF12, SPGF15
UniGene
Hs.553795
Gene Chromosome Location
Chr.10: 133553900-133568291 on Build GRCh38
Transcripts
NM_001143763.1 NM_001143764.1 NM_130784.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5017929 c.273G>C p.L91L Chr.10: 133558213-133558213 on Build GRCh38
1703300 c.169G>A p.G57R Chr.10: 133558209-133558209 on Build GRCh38
5370407 c.174G>A p.E58E Chr.10: 133558204-133558204 on Build GRCh38
1703299 c.277G>A p.G93R Chr.10: 133558209-133558209 on Build GRCh38
5017930 c.165G>C p.L55L Chr.10: 133558213-133558213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894051647 Chr.10: 133558164-133558164 on Build GRCh38
rs768730141 Chr.10: 133558116-133558116 on Build GRCh38
rs759048457 Chr.10: 133558145-133558145 on Build GRCh38
rs371072591 Chr.10: 133558177-133558177 on Build GRCh38
rs778894333 Chr.10: 133558183-133558183 on Build GRCh38
rs772445757 Chr.10: 133558126-133558126 on Build GRCh38
rs199850383 Chr.10: 133558156-133558156 on Build GRCh38
rs116399728 Chr.10: 133558157-133558157 on Build GRCh38
rs773413090 Chr.10: 133558128-133558128 on Build GRCh38
rs762748967 Chr.10: 133558161-133558161 on Build GRCh38
rs564389538 Chr.10: 133558099-133558099 on Build GRCh38
rs748296472 Chr.10: 133558190-133558190 on Build GRCh38
rs181979224 Chr.10: 133558263-133558263 on Build GRCh38
rs765142984 Chr.10: 133558133-133558133 on Build GRCh38
rs947789758 Chr.10: 133558135-133558135 on Build GRCh38
rs772432370 Chr.10: 133558195-133558195 on Build GRCh38
rs1039561352 Chr.10: 133558176-133558176 on Build GRCh38
rs756799473 Chr.10: 133558256-133558257 on Build GRCh38
rs751457315 Chr.10: 133558228-133558229 on Build GRCh38
rs780185568 Chr.10: 133558166-133558166 on Build GRCh38
rs776490257 Chr.10: 133558140-133558140 on Build GRCh38
rs755100 Chr.10: 133558121-133558121 on Build GRCh38
rs8181458 Chr.10: 133558102-133558102 on Build GRCh38
rs766114034 Chr.10: 133558138-133558138 on Build GRCh38
rs771424279 Chr.10: 133558239-133558239 on Build GRCh38
rs775149425 Chr.10: 133558150-133558151 on Build GRCh38
rs368519924 Chr.10: 133558158-133558158 on Build GRCh38
rs780118418 Chr.10: 133558134-133558134 on Build GRCh38
rs747372056 Chr.10: 133558238-133558238 on Build GRCh38
rs764666015 Chr.10: 133558152-133558152 on Build GRCh38
rs575617775 Chr.10: 133558210-133558210 on Build GRCh38
rs199922141 Chr.10: 133558109-133558109 on Build GRCh38
rs763958481 Chr.10: 133558170-133558171 on Build GRCh38
rs752246204 Chr.10: 133558273-133558273 on Build GRCh38
rs752630923 Chr.10: 133558154-133558154 on Build GRCh38
rs761166425 Chr.10: 133558133-133558133 on Build GRCh38
rs751993409 Chr.10: 133558141-133558141 on Build GRCh38
rs754075575 Chr.10: 133558167-133558167 on Build GRCh38
rs748628823 Chr.10: 133558125-133558125 on Build GRCh38

View Full Product Details