Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119185746-119185995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD3
Entrez Gene ID
79849
Gene Name
PDZ domain containing 3
Gene Aliases
IKEPP, NHERF4, PDZK2
UniGene
Hs.374726
Gene Chromosome Location
Chr.11: 119185457-119190223 on Build GRCh38
Transcripts
NM_001168468.1 NM_024791.3 NR_033122.1 XM_011543000.2 XM_011543001.2 XM_011543002.2 XM_011543005.2 XM_017018334.1 XM_017018335.1 XM_017018336.1 XM_017018337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5435862 c.29C>T p.T10I Chr.11: 119185937-119185937 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs184359193 Chr.11: 119185963-119185963 on Build GRCh38
rs557483583 Chr.11: 119185882-119185883 on Build GRCh38
rs138112455 Chr.11: 119185936-119185936 on Build GRCh38
rs933124717 Chr.11: 119185779-119185779 on Build GRCh38
rs920315487 Chr.11: 119185973-119185973 on Build GRCh38
rs904158554 Chr.11: 119185852-119185852 on Build GRCh38
rs749936614 Chr.11: 119185875-119185875 on Build GRCh38
rs540081421 Chr.11: 119185781-119185781 on Build GRCh38
rs769591612 Chr.11: 119185953-119185954 on Build GRCh38
rs772694760 Chr.11: 119185944-119185944 on Build GRCh38
rs762498139 Chr.11: 119185960-119185960 on Build GRCh38
rs777923995 Chr.11: 119185833-119185833 on Build GRCh38
rs745734182 Chr.11: 119185839-119185839 on Build GRCh38
rs749439728 Chr.11: 119185906-119185906 on Build GRCh38
rs761657006 Chr.11: 119185847-119185847 on Build GRCh38
rs532344731 Chr.11: 119185788-119185788 on Build GRCh38
rs776104211 Chr.11: 119185867-119185867 on Build GRCh38
rs775414070 Chr.11: 119185853-119185853 on Build GRCh38
rs371843707 Chr.11: 119185892-119185892 on Build GRCh38
rs1041325929 Chr.11: 119185815-119185815 on Build GRCh38
rs748433865 Chr.11: 119185814-119185814 on Build GRCh38
rs1031492888 Chr.11: 119185907-119185907 on Build GRCh38
rs768327702 Chr.11: 119185861-119185861 on Build GRCh38
rs1007429194 Chr.11: 119185812-119185812 on Build GRCh38
rs748833685 Chr.11: 119185952-119185953 on Build GRCh38
rs45612531 Chr.11: 119185972-119185972 on Build GRCh38
rs780184915 Chr.11: 119185909-119185909 on Build GRCh38
rs761545329 Chr.11: 119185868-119185868 on Build GRCh38
rs781637056 Chr.11: 119185898-119185898 on Build GRCh38
rs776181588 Chr.11: 119185893-119185895 on Build GRCh38
rs752806323 Chr.11: 119185902-119185902 on Build GRCh38
rs769554452 Chr.11: 119185943-119185943 on Build GRCh38
rs191882344 Chr.11: 119185912-119185912 on Build GRCh38
rs772206646 Chr.11: 119185842-119185842 on Build GRCh38
rs763444293 Chr.11: 119185886-119185886 on Build GRCh38
rs562744717 Chr.11: 119185903-119185903 on Build GRCh38
rs776495808 Chr.11: 119185929-119185929 on Build GRCh38
rs200186795 Chr.11: 119185964-119185964 on Build GRCh38
rs899821378 Chr.11: 119185846-119185846 on Build GRCh38
rs761409369 Chr.11: 119185909-119185911 on Build GRCh38
rs1025143691 Chr.11: 119185921-119185921 on Build GRCh38
rs565086576 Chr.11: 119185784-119185784 on Build GRCh38
rs550902994 Chr.11: 119185871-119185871 on Build GRCh38
rs746995004 Chr.11: 119185910-119185910 on Build GRCh38
rs759663473 Chr.11: 119185774-119185778 on Build GRCh38
rs751899069 Chr.11: 119185888-119185888 on Build GRCh38
rs552538682 Chr.11: 119185969-119185969 on Build GRCh38
rs970090320 Chr.11: 119185966-119185966 on Build GRCh38
rs769714066 Chr.11: 119185821-119185821 on Build GRCh38

View Full Product Details