Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67675374-67675647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928045
Entrez Gene ID
101928045
Gene Name
uncharacterized LOC101928045
Gene Aliases
-
UniGene
Hs.231295
Gene Chromosome Location
Chr.17: 67657206-67690871 on Build GRCh38
Transcripts
NM_012417.3 NM_181671.2 XM_005257216.2 XM_017024442.1 XM_017024443.1 XM_017024444.1 XM_017024445.1

Gene Symbol
PITPNC1
Entrez Gene ID
26207
Gene Name
phosphatidylinositol transfer protein, cytoplasmic 1
Gene Aliases
M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1
UniGene
Hs.591185
Gene Chromosome Location
Chr.17: 67377281-67697263 on Build GRCh38
Transcripts
NM_012417.3 NM_181671.2 XM_005257216.2 XM_017024442.1 XM_017024443.1 XM_017024444.1 XM_017024445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972361 c.663A>G p.A221A Chr.17: 67675523-67675523 on Build GRCh38
4817257 c.670G>A p.D224N Chr.17: 67675530-67675530 on Build GRCh38
2797543 c.624C>T p.V208V Chr.17: 67675484-67675484 on Build GRCh38
5972359 c.663A>G p.A221A Chr.17: 67675523-67675523 on Build GRCh38
6016785 c.625C>T p.R209* Chr.17: 67675485-67675485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531778964 Chr.17: 67675614-67675614 on Build GRCh38
rs749009010 Chr.17: 67675570-67675570 on Build GRCh38
rs757546900 Chr.17: 67675486-67675486 on Build GRCh38
rs1049008639 Chr.17: 67675529-67675529 on Build GRCh38
rs745395827 Chr.17: 67675587-67675587 on Build GRCh38
rs761033711 Chr.17: 67675442-67675442 on Build GRCh38
rs773982743 Chr.17: 67675473-67675473 on Build GRCh38
rs774978384 Chr.17: 67675592-67675592 on Build GRCh38
rs1056473386 Chr.17: 67675609-67675609 on Build GRCh38
rs753144727 Chr.17: 67675437-67675437 on Build GRCh38
rs992983940 Chr.17: 67675424-67675424 on Build GRCh38
rs754137801 Chr.17: 67675459-67675459 on Build GRCh38
rs778971773 Chr.17: 67675501-67675501 on Build GRCh38
rs781285228 Chr.17: 67675554-67675554 on Build GRCh38
rs781732841 Chr.17: 67675560-67675560 on Build GRCh38
rs771504300 Chr.17: 67675591-67675591 on Build GRCh38
rs758498976 Chr.17: 67675553-67675553 on Build GRCh38
rs764588021 Chr.17: 67675458-67675458 on Build GRCh38
rs921775644 Chr.17: 67675448-67675448 on Build GRCh38
rs561150478 Chr.17: 67675541-67675541 on Build GRCh38
rs369474786 Chr.17: 67675569-67675569 on Build GRCh38
rs36114934 Chr.17: 67675517-67675517 on Build GRCh38
rs773831635 Chr.17: 67675585-67675585 on Build GRCh38
rs917312713 Chr.17: 67675533-67675533 on Build GRCh38
rs768038618 Chr.17: 67675430-67675430 on Build GRCh38

View Full Product Details