Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41688705-41688934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNU13
Entrez Gene ID
4809
Gene Name
SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5)
Gene Aliases
15.5K, FA-1, FA1, NHP2L1, NHPX, OTK27, SNRNP15-5, SPAG12, SSFA1
UniGene
Hs.182255
Gene Chromosome Location
Chr.22: 41673933-41690492 on Build GRCh38
Transcripts
NM_001003796.1 NM_005008.3 XM_017028811.1

SNP Detail

SNP ID Chromosome Location
rs967327649 Chr.22: 41688896-41688896 on Build GRCh38
rs770654443 Chr.22: 41688746-41688746 on Build GRCh38
rs747796031 Chr.22: 41688869-41688869 on Build GRCh38
rs762194025 Chr.22: 41688838-41688838 on Build GRCh38
rs370244196 Chr.22: 41688767-41688767 on Build GRCh38
rs182469753 Chr.22: 41688916-41688916 on Build GRCh38
rs929007009 Chr.22: 41688876-41688877 on Build GRCh38
rs750857288 Chr.22: 41688782-41688782 on Build GRCh38
rs998663074 Chr.22: 41688803-41688803 on Build GRCh38
rs755594812 Chr.22: 41688808-41688808 on Build GRCh38
rs539240871 Chr.22: 41688809-41688809 on Build GRCh38
rs199776499 Chr.22: 41688830-41688830 on Build GRCh38
rs752185586 Chr.22: 41688802-41688802 on Build GRCh38
rs370253652 Chr.22: 41688823-41688823 on Build GRCh38
rs764591965 Chr.22: 41688773-41688773 on Build GRCh38
rs779016339 Chr.22: 41688779-41688780 on Build GRCh38
rs765727513 Chr.22: 41688779-41688779 on Build GRCh38
rs748215313 Chr.22: 41688817-41688818 on Build GRCh38
rs542091245 Chr.22: 41688876-41688876 on Build GRCh38
rs775537014 Chr.22: 41688771-41688771 on Build GRCh38
rs867224102 Chr.22: 41688777-41688777 on Build GRCh38
rs543374446 Chr.22: 41688886-41688886 on Build GRCh38
rs770471718 Chr.22: 41688841-41688841 on Build GRCh38
rs768636927 Chr.22: 41688834-41688834 on Build GRCh38
rs745722505 Chr.22: 41688829-41688829 on Build GRCh38
rs557567384 Chr.22: 41688865-41688865 on Build GRCh38
rs777151557 Chr.22: 41688837-41688837 on Build GRCh38
rs760731624 Chr.22: 41688772-41688772 on Build GRCh38
rs757228709 Chr.22: 41688818-41688818 on Build GRCh38
rs747050365 Chr.22: 41688832-41688832 on Build GRCh38
rs940626353 Chr.22: 41688750-41688750 on Build GRCh38
rs767335368 Chr.22: 41688796-41688796 on Build GRCh38
rs1022425119 Chr.22: 41688877-41688877 on Build GRCh38
rs1028387060 Chr.22: 41688825-41688825 on Build GRCh38
rs749215482 Chr.22: 41688811-41688811 on Build GRCh38
rs746062555 Chr.22: 41688764-41688764 on Build GRCh38
rs201752163 Chr.22: 41688786-41688786 on Build GRCh38
rs774520657 Chr.22: 41688747-41688747 on Build GRCh38
rs978387811 Chr.22: 41688903-41688903 on Build GRCh38
rs777013490 Chr.22: 41688774-41688774 on Build GRCh38
rs1035072505 Chr.22: 41688910-41688910 on Build GRCh38
rs887474958 Chr.22: 41688887-41688887 on Build GRCh38
rs779805144 Chr.22: 41688749-41688749 on Build GRCh38

View Full Product Details