Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204117528-204117793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOX13
Entrez Gene ID
9580
Gene Name
SRY-box 13
Gene Aliases
ICA12, Sox-13
UniGene
Hs.201671
Gene Chromosome Location
Chr.1: 204073096-204127743 on Build GRCh38
Transcripts
NM_005686.2 XM_005245623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5899237 c.764C>T p.P255L Chr.1: 204117696-204117696 on Build GRCh38
902553 c.687C>A p.I229I Chr.1: 204117619-204117619 on Build GRCh38
6541601 c.750C>T p.P250P Chr.1: 204117682-204117682 on Build GRCh38
4847440 c.715C>G p.L239V Chr.1: 204117647-204117647 on Build GRCh38
4505569 c.696C>T p.F232F Chr.1: 204117628-204117628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779167472 Chr.1: 204117631-204117631 on Build GRCh38
rs764727612 Chr.1: 204117565-204117565 on Build GRCh38
rs369370682 Chr.1: 204117603-204117603 on Build GRCh38
rs767222195 Chr.1: 204117738-204117738 on Build GRCh38
rs561788252 Chr.1: 204117619-204117619 on Build GRCh38
rs1002332744 Chr.1: 204117730-204117730 on Build GRCh38
rs761067750 Chr.1: 204117556-204117556 on Build GRCh38
rs185065841 Chr.1: 204117739-204117739 on Build GRCh38
rs763875353 Chr.1: 204117705-204117705 on Build GRCh38
rs890426173 Chr.1: 204117726-204117726 on Build GRCh38
rs553115254 Chr.1: 204117558-204117558 on Build GRCh38
rs191743243 Chr.1: 204117674-204117674 on Build GRCh38
rs148336151 Chr.1: 204117554-204117554 on Build GRCh38
rs541906019 Chr.1: 204117564-204117564 on Build GRCh38
rs1032116613 Chr.1: 204117771-204117771 on Build GRCh38
rs1043928280 Chr.1: 204117718-204117718 on Build GRCh38
rs751410553 Chr.1: 204117610-204117610 on Build GRCh38
rs758675851 Chr.1: 204117759-204117759 on Build GRCh38
rs375834163 Chr.1: 204117601-204117601 on Build GRCh38
rs757138043 Chr.1: 204117613-204117613 on Build GRCh38
rs748281908 Chr.1: 204117756-204117756 on Build GRCh38
rs376489680 Chr.1: 204117573-204117573 on Build GRCh38
rs200747952 Chr.1: 204117664-204117664 on Build GRCh38
rs1037790958 Chr.1: 204117635-204117635 on Build GRCh38
rs904002846 Chr.1: 204117720-204117720 on Build GRCh38
rs751298991 Chr.1: 204117731-204117731 on Build GRCh38
rs201294252 Chr.1: 204117755-204117755 on Build GRCh38
rs896538934 Chr.1: 204117741-204117741 on Build GRCh38
rs771210642 Chr.1: 204117644-204117644 on Build GRCh38
rs769691561 Chr.1: 204117753-204117753 on Build GRCh38
rs987805805 Chr.1: 204117555-204117555 on Build GRCh38
rs57825158 Chr.1: 204117560-204117560 on Build GRCh38
rs758066639 Chr.1: 204117587-204117587 on Build GRCh38
rs141412155 Chr.1: 204117768-204117768 on Build GRCh38
rs564205881 Chr.1: 204117746-204117746 on Build GRCh38
rs768790788 Chr.1: 204117626-204117626 on Build GRCh38
rs761155084 Chr.1: 204117633-204117633 on Build GRCh38
rs748044555 Chr.1: 204117611-204117611 on Build GRCh38
rs376348030 Chr.1: 204117650-204117650 on Build GRCh38
rs777408216 Chr.1: 204117609-204117609 on Build GRCh38
rs544550219 Chr.1: 204117682-204117682 on Build GRCh38
rs36055883 Chr.1: 204117632-204117632 on Build GRCh38
rs776685914 Chr.1: 204117645-204117646 on Build GRCh38
rs764676220 Chr.1: 204117666-204117666 on Build GRCh38
rs745808614 Chr.1: 204117625-204117625 on Build GRCh38
rs974894934 Chr.1: 204117577-204117577 on Build GRCh38
rs369807881 Chr.1: 204117732-204117732 on Build GRCh38

View Full Product Details