Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113273109-113273382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPCN1
Entrez Gene ID
53373
Gene Name
two pore segment channel 1
Gene Aliases
TPC1
UniGene
Hs.524763
Gene Chromosome Location
Chr.12: 113221429-113298589 on Build GRCh38
Transcripts
NM_001143819.2 NM_001301214.1 NM_017901.5 XM_005253906.4 XM_011538490.2 XM_011538492.2 XM_011538493.2 XM_017019480.1 XM_017019481.1 XM_017019482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6806737 c.788T>C p.F263S Chr.12: 113273236-113273236 on Build GRCh38
1746764 c.1005C>T p.F335F Chr.12: 113273237-113273237 on Build GRCh38
6806736 c.1004T>C p.F335S Chr.12: 113273236-113273236 on Build GRCh38
242015 c.811G>A p.V271I Chr.12: 113273259-113273259 on Build GRCh38
6806733 c.784T>C p.Y262H Chr.12: 113273232-113273232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751296676 Chr.12: 113273255-113273255 on Build GRCh38
rs192329797 Chr.12: 113273201-113273201 on Build GRCh38
rs767287535 Chr.12: 113273267-113273267 on Build GRCh38
rs759535132 Chr.12: 113273132-113273132 on Build GRCh38
rs557584323 Chr.12: 113273239-113273239 on Build GRCh38
rs777522895 Chr.12: 113273185-113273185 on Build GRCh38
rs776776956 Chr.12: 113273203-113273203 on Build GRCh38
rs752124612 Chr.12: 113273307-113273307 on Build GRCh38
rs112480087 Chr.12: 113273198-113273198 on Build GRCh38
rs984122185 Chr.12: 113273296-113273296 on Build GRCh38
rs754314089 Chr.12: 113273344-113273344 on Build GRCh38
rs369275415 Chr.12: 113273183-113273183 on Build GRCh38
rs765466463 Chr.12: 113273218-113273218 on Build GRCh38
rs370205106 Chr.12: 113273326-113273326 on Build GRCh38
rs752749864 Chr.12: 113273274-113273276 on Build GRCh38
rs761304174 Chr.12: 113273200-113273200 on Build GRCh38
rs6489894 Chr.12: 113273143-113273143 on Build GRCh38
rs756959450 Chr.12: 113273330-113273330 on Build GRCh38
rs755738113 Chr.12: 113273317-113273317 on Build GRCh38
rs763129522 Chr.12: 113273228-113273228 on Build GRCh38
rs948098473 Chr.12: 113273341-113273341 on Build GRCh38
rs151327811 Chr.12: 113273186-113273186 on Build GRCh38
rs772608112 Chr.12: 113273189-113273189 on Build GRCh38
rs778335040 Chr.12: 113273342-113273345 on Build GRCh38
rs201637566 Chr.12: 113273204-113273204 on Build GRCh38
rs761307152 Chr.12: 113273323-113273323 on Build GRCh38
rs182098432 Chr.12: 113273331-113273331 on Build GRCh38
rs756668928 Chr.12: 113273317-113273317 on Build GRCh38
rs11066524 Chr.12: 113273291-113273291 on Build GRCh38
rs754953269 Chr.12: 113273258-113273258 on Build GRCh38
rs573352135 Chr.12: 113273301-113273301 on Build GRCh38
rs747551420 Chr.12: 113273182-113273182 on Build GRCh38

View Full Product Details