Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94910332-94910591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R9A
Entrez Gene ID
55607
Gene Name
protein phosphatase 1 regulatory subunit 9A
Gene Aliases
NRB1, NRBI, Neurabin-I
UniGene
Hs.21816
Gene Chromosome Location
Chr.7: 94907202-95296415 on Build GRCh38
Transcripts
NM_001166160.1 NM_001166161.1 NM_001166162.1 NM_001166163.1 NM_017650.2 XM_011516380.1 XM_011516381.1 XM_011516382.2 XM_011516383.1 XM_011516389.1 XM_017012394.1 XM_017012395.1 XM_017012396.1 XM_017012397.1 XM_017012398.1 XM_017012399.1 XM_017012400.1 XM_017012401.1 XM_017012402.1 XM_017012403.1 XM_017012404.1 XM_017012405.1 XM_017012406.1 XM_017012407.1 XM_017012409.1 XM_017012410.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6754170 c.363T>C p.S121S Chr.7: 94910476-94910476 on Build GRCh38
3942393 c.356C>T p.S119F Chr.7: 94910469-94910469 on Build GRCh38
1496707 c.279T>C p.H93H Chr.7: 94910392-94910392 on Build GRCh38
1452998 c.263delG p.G91fs*10 Chr.7: 94910376-94910376 on Build GRCh38
4486581 c.306C>T p.P102P Chr.7: 94910419-94910419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758470260 Chr.7: 94910544-94910544 on Build GRCh38
rs760073246 Chr.7: 94910442-94910442 on Build GRCh38
rs866642324 Chr.7: 94910380-94910380 on Build GRCh38
rs781472900 Chr.7: 94910372-94910372 on Build GRCh38
rs993310945 Chr.7: 94910366-94910366 on Build GRCh38
rs772763566 Chr.7: 94910397-94910399 on Build GRCh38
rs958939237 Chr.7: 94910360-94910360 on Build GRCh38
rs1039634547 Chr.7: 94910552-94910552 on Build GRCh38
rs987070252 Chr.7: 94910421-94910421 on Build GRCh38
rs930070751 Chr.7: 94910418-94910418 on Build GRCh38
rs375944580 Chr.7: 94910536-94910536 on Build GRCh38
rs769981262 Chr.7: 94910388-94910388 on Build GRCh38
rs941242228 Chr.7: 94910540-94910540 on Build GRCh38
rs1047023936 Chr.7: 94910458-94910458 on Build GRCh38
rs753550890 Chr.7: 94910507-94910507 on Build GRCh38
rs773491216 Chr.7: 94910407-94910407 on Build GRCh38
rs775037733 Chr.7: 94910470-94910470 on Build GRCh38
rs771345715 Chr.7: 94910416-94910416 on Build GRCh38
rs750366511 Chr.7: 94910524-94910524 on Build GRCh38
rs917364633 Chr.7: 94910409-94910409 on Build GRCh38
rs768827470 Chr.7: 94910361-94910361 on Build GRCh38
rs781419894 Chr.7: 94910547-94910547 on Build GRCh38
rs777531041 Chr.7: 94910435-94910435 on Build GRCh38
rs765122221 Chr.7: 94910510-94910510 on Build GRCh38
rs748369962 Chr.7: 94910384-94910384 on Build GRCh38
rs774677192 Chr.7: 94910423-94910423 on Build GRCh38
rs368743894 Chr.7: 94910463-94910463 on Build GRCh38
rs374284975 Chr.7: 94910413-94910413 on Build GRCh38
rs370361768 Chr.7: 94910506-94910506 on Build GRCh38
rs773851914 Chr.7: 94910456-94910456 on Build GRCh38
rs554560312 Chr.7: 94910498-94910498 on Build GRCh38

View Full Product Details