Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61445880-61446135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHAF2
Entrez Gene ID
54949
Gene Name
succinate dehydrogenase complex assembly factor 2
Gene Aliases
C11orf79, PGL2, SDH5
UniGene
Hs.313247
Gene Chromosome Location
Chr.11: 61430125-61446767 on Build GRCh38
Transcripts
NM_017841.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7147667 c.475G>C p.E159Q Chr.11: 61446045-61446045 on Build GRCh38
7150753 c.420G>A p.L140L Chr.11: 61445990-61445990 on Build GRCh38
6483097 c.453G>C p.Q151H Chr.11: 61446023-61446023 on Build GRCh38
3869707 c.466C>T p.P156S Chr.11: 61446036-61446036 on Build GRCh38
7266178 c.411G>A p.M137I Chr.11: 61445981-61445981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759780830 Chr.11: 61445991-61445991 on Build GRCh38
rs755233768 Chr.11: 61445997-61445997 on Build GRCh38
rs758154733 Chr.11: 61446035-61446035 on Build GRCh38
rs774723481 Chr.11: 61445990-61445990 on Build GRCh38
rs775160119 Chr.11: 61445937-61445937 on Build GRCh38
rs113652589 Chr.11: 61446083-61446083 on Build GRCh38
rs772219112 Chr.11: 61446015-61446016 on Build GRCh38
rs776140713 Chr.11: 61446068-61446068 on Build GRCh38
rs773580529 Chr.11: 61445961-61445961 on Build GRCh38
rs757151603 Chr.11: 61446029-61446029 on Build GRCh38
rs778585796 Chr.11: 61446030-61446030 on Build GRCh38
rs759866664 Chr.11: 61446091-61446091 on Build GRCh38
rs964701048 Chr.11: 61446087-61446087 on Build GRCh38
rs1006514566 Chr.11: 61445952-61445952 on Build GRCh38
rs370528677 Chr.11: 61445920-61445920 on Build GRCh38
rs572576746 Chr.11: 61446000-61446000 on Build GRCh38
rs753711279 Chr.11: 61446028-61446028 on Build GRCh38
rs993562199 Chr.11: 61445913-61445913 on Build GRCh38
rs768048172 Chr.11: 61446067-61446067 on Build GRCh38
rs749776601 Chr.11: 61446080-61446080 on Build GRCh38
rs775673914 Chr.11: 61446094-61446094 on Build GRCh38
rs756162021 Chr.11: 61446009-61446009 on Build GRCh38
rs1002286076 Chr.11: 61446009-61446009 on Build GRCh38
rs367574730 Chr.11: 61445980-61445980 on Build GRCh38
rs764146390 Chr.11: 61446020-61446020 on Build GRCh38
rs770383127 Chr.11: 61445957-61445957 on Build GRCh38
rs1056273933 Chr.11: 61446024-61446024 on Build GRCh38
rs373096675 Chr.11: 61445949-61445949 on Build GRCh38
rs771792041 Chr.11: 61445929-61445929 on Build GRCh38
rs375280597 Chr.11: 61445939-61445939 on Build GRCh38
rs114594435 Chr.11: 61446104-61446104 on Build GRCh38
rs773301877 Chr.11: 61446058-61446059 on Build GRCh38
rs370174263 Chr.11: 61446021-61446021 on Build GRCh38
rs766658781 Chr.11: 61445984-61445984 on Build GRCh38
rs538679827 Chr.11: 61445905-61445905 on Build GRCh38
rs150187184 Chr.11: 61446060-61446060 on Build GRCh38
rs113028338 Chr.11: 61446079-61446079 on Build GRCh38
rs768954974 Chr.11: 61446031-61446031 on Build GRCh38
rs560146405 Chr.11: 61446084-61446084 on Build GRCh38
rs760871747 Chr.11: 61446098-61446098 on Build GRCh38
rs767772474 Chr.11: 61446093-61446093 on Build GRCh38
rs968303021 Chr.11: 61446066-61446066 on Build GRCh38
rs140920079 Chr.11: 61446046-61446046 on Build GRCh38

View Full Product Details