Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185323489-185323732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX25
Entrez Gene ID
83891
Gene Name
sorting nexin 25
Gene Aliases
MSTP043, SBBI31
UniGene
Hs.369091
Gene Chromosome Location
Chr.4: 185204237-185363971 on Build GRCh38
Transcripts
NM_001317781.1 XM_005263267.4 XM_011532302.2 XM_011532303.2 XM_011532304.2 XM_011532305.2 XM_011532306.2 XM_011532307.2 XM_011532308.2 XM_017008680.1 XM_017008681.1 XM_017008682.1 XM_017008683.1 XM_017008684.1 XM_017008685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419432 c.1156A>G p.R386G Chr.4: 185323699-185323699 on Build GRCh38
4123726 c.1049C>A p.S350Y Chr.4: 185323592-185323592 on Build GRCh38
4135520 c.1152G>A p.K384K Chr.4: 185323695-185323695 on Build GRCh38
1053766 c.1042G>T p.E348* Chr.4: 185323585-185323585 on Build GRCh38
1053767 c.1171T>C p.F391L Chr.4: 185323714-185323714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987756318 Chr.4: 185323649-185323649 on Build GRCh38
rs78573153 Chr.4: 185323575-185323575 on Build GRCh38
rs759864394 Chr.4: 185323557-185323557 on Build GRCh38
rs747047316 Chr.4: 185323549-185323549 on Build GRCh38
rs771301551 Chr.4: 185323597-185323597 on Build GRCh38
rs148867014 Chr.4: 185323533-185323533 on Build GRCh38
rs749435584 Chr.4: 185323661-185323661 on Build GRCh38
rs745513354 Chr.4: 185323700-185323700 on Build GRCh38
rs773796826 Chr.4: 185323530-185323530 on Build GRCh38
rs767059444 Chr.4: 185323542-185323542 on Build GRCh38
rs985194364 Chr.4: 185323559-185323559 on Build GRCh38
rs755597794 Chr.4: 185323591-185323591 on Build GRCh38
rs781436636 Chr.4: 185323633-185323633 on Build GRCh38
rs752319958 Chr.4: 185323588-185323588 on Build GRCh38
rs927106911 Chr.4: 185323662-185323662 on Build GRCh38
rs553227310 Chr.4: 185323652-185323652 on Build GRCh38
rs1057187788 Chr.4: 185323704-185323704 on Build GRCh38
rs1029202440 Chr.4: 185323526-185323526 on Build GRCh38
rs748340775 Chr.4: 185323635-185323635 on Build GRCh38
rs35748792 Chr.4: 185323608-185323608 on Build GRCh38
rs567509062 Chr.4: 185323604-185323604 on Build GRCh38
rs1042866833 Chr.4: 185323659-185323659 on Build GRCh38
rs771540400 Chr.4: 185323694-185323694 on Build GRCh38
rs769629651 Chr.4: 185323701-185323701 on Build GRCh38
rs36102675 Chr.4: 185323690-185323690 on Build GRCh38
rs371526877 Chr.4: 185323617-185323617 on Build GRCh38
rs550724396 Chr.4: 185323602-185323602 on Build GRCh38
rs751957546 Chr.4: 185323544-185323544 on Build GRCh38
rs3733643 Chr.4: 185323539-185323539 on Build GRCh38
rs777282679 Chr.4: 185323595-185323595 on Build GRCh38
rs769845347 Chr.4: 185323637-185323637 on Build GRCh38
rs758169639 Chr.4: 185323621-185323621 on Build GRCh38
rs17852713 Chr.4: 185323698-185323698 on Build GRCh38
rs773420372 Chr.4: 185323651-185323651 on Build GRCh38

View Full Product Details