Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65625047-65625274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC24A1
Entrez Gene ID
9187
Gene Name
solute carrier family 24 member 1
Gene Aliases
CSNB1D, HsT17412, NCKX, NCKX1, RODX
UniGene
Hs.173092
Gene Chromosome Location
Chr.15: 65611405-65660995 on Build GRCh38
Transcripts
NM_001301031.1 NM_001301032.1 NM_001301033.1 NM_004727.2 XM_005254778.3 XM_005254781.3 XM_006720768.3 XM_011522219.1 XM_011522220.2 XM_011522221.2 XM_011522222.2 XM_011522226.2 XM_017022724.1 XM_017022725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848582 c.1033C>G p.L345V Chr.15: 65625113-65625113 on Build GRCh38
964091 c.1009A>G p.K337E Chr.15: 65625089-65625089 on Build GRCh38
6332590 c.1139C>T p.T380I Chr.15: 65625219-65625219 on Build GRCh38
1129141 c.1062A>G p.V354V Chr.15: 65625142-65625142 on Build GRCh38
6996826 c.989G>A p.G330D Chr.15: 65625069-65625069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189424576 Chr.15: 65625139-65625139 on Build GRCh38
rs755663087 Chr.15: 65625202-65625202 on Build GRCh38
rs764426619 Chr.15: 65625122-65625122 on Build GRCh38
rs775976233 Chr.15: 65625214-65625214 on Build GRCh38
rs937377731 Chr.15: 65625133-65625133 on Build GRCh38
rs769943891 Chr.15: 65625226-65625226 on Build GRCh38
rs377117199 Chr.15: 65625251-65625251 on Build GRCh38
rs761002618 Chr.15: 65625119-65625119 on Build GRCh38
rs750376695 Chr.15: 65625158-65625158 on Build GRCh38
rs375552552 Chr.15: 65625174-65625174 on Build GRCh38
rs766181605 Chr.15: 65625172-65625172 on Build GRCh38
rs754554767 Chr.15: 65625182-65625182 on Build GRCh38
rs765076828 Chr.15: 65625153-65625153 on Build GRCh38
rs747598530 Chr.15: 65625191-65625191 on Build GRCh38
rs1032145376 Chr.15: 65625099-65625099 on Build GRCh38
rs372351507 Chr.15: 65625113-65625113 on Build GRCh38
rs374623693 Chr.15: 65625215-65625215 on Build GRCh38
rs373518306 Chr.15: 65625076-65625077 on Build GRCh38
rs779497185 Chr.15: 65625210-65625210 on Build GRCh38
rs368902751 Chr.15: 65625246-65625246 on Build GRCh38
rs574082600 Chr.15: 65625071-65625071 on Build GRCh38
rs199504782 Chr.15: 65625211-65625211 on Build GRCh38
rs780962971 Chr.15: 65625187-65625187 on Build GRCh38
rs139200508 Chr.15: 65625225-65625225 on Build GRCh38
rs776544893 Chr.15: 65625124-65625124 on Build GRCh38
rs776973720 Chr.15: 65625216-65625216 on Build GRCh38
rs553194871 Chr.15: 65625170-65625170 on Build GRCh38
rs954904049 Chr.15: 65625238-65625238 on Build GRCh38

View Full Product Details