Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72130003-72130276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
-
UniGene
Hs.397836
Gene Chromosome Location
Chr.X: 71911073-72153286 on Build GRCh38
Transcripts
NM_001013627.2 XM_011530932.1 XM_011530933.1 XM_011530934.2 XM_017029473.1 XM_017029474.1 XM_017029475.1 XM_017029476.1 XM_017029477.1 NM_001024455.3

Gene Symbol
RGAG4
Entrez Gene ID
340526
Gene Name
retrotransposon gag domain containing 4
Gene Aliases
6430402L03Rik, MAR5, MART5
UniGene
Hs.512180
Gene Chromosome Location
Chr.X: 72127110-72131901 on Build GRCh38
Transcripts
NM_001013627.2 XM_011530932.1 XM_011530933.1 XM_011530934.2 XM_017029473.1 XM_017029474.1 XM_017029475.1 XM_017029476.1 XM_017029477.1 NM_001024455.3

SNP Detail

SNP ID Chromosome Location
rs755133990 Chr.X: 72130059-72130059 on Build GRCh38
rs764230784 Chr.X: 72130172-72130172 on Build GRCh38
rs781715793 Chr.X: 72130078-72130078 on Build GRCh38
rs757021429 Chr.X: 72130125-72130125 on Build GRCh38
rs780558080 Chr.X: 72130187-72130187 on Build GRCh38
rs1028313139 Chr.X: 72130025-72130025 on Build GRCh38
rs372733180 Chr.X: 72130113-72130113 on Build GRCh38
rs777354507 Chr.X: 72130205-72130205 on Build GRCh38
rs367678202 Chr.X: 72130198-72130198 on Build GRCh38
rs778358543 Chr.X: 72130111-72130111 on Build GRCh38
rs759517559 Chr.X: 72130135-72130135 on Build GRCh38
rs763116118 Chr.X: 72130071-72130071 on Build GRCh38
rs750913102 Chr.X: 72130092-72130092 on Build GRCh38
rs771005184 Chr.X: 72130037-72130037 on Build GRCh38
rs770892372 Chr.X: 72130226-72130226 on Build GRCh38
rs370818026 Chr.X: 72130070-72130070 on Build GRCh38
rs759755785 Chr.X: 72130055-72130055 on Build GRCh38
rs756640557 Chr.X: 72130186-72130186 on Build GRCh38
rs987051099 Chr.X: 72130096-72130096 on Build GRCh38
rs60726595 Chr.X: 72130143-72130143 on Build GRCh38
rs771936813 Chr.X: 72130024-72130024 on Build GRCh38
rs760468437 Chr.X: 72130034-72130034 on Build GRCh38
rs752160610 Chr.X: 72130189-72130189 on Build GRCh38
rs762226176 Chr.X: 72130175-72130175 on Build GRCh38
rs753625204 Chr.X: 72130112-72130112 on Build GRCh38
rs767842591 Chr.X: 72130183-72130183 on Build GRCh38
rs200189694 Chr.X: 72130073-72130073 on Build GRCh38
rs776759820 Chr.X: 72130128-72130128 on Build GRCh38
rs201171574 Chr.X: 72130110-72130110 on Build GRCh38
rs750966235 Chr.X: 72130185-72130185 on Build GRCh38
rs776641896 Chr.X: 72130049-72130049 on Build GRCh38
rs775454517 Chr.X: 72130151-72130151 on Build GRCh38
rs995909319 Chr.X: 72130127-72130127 on Build GRCh38
rs770843646 Chr.X: 72130126-72130126 on Build GRCh38
rs781166664 Chr.X: 72130231-72130231 on Build GRCh38
rs745766699 Chr.X: 72130169-72130169 on Build GRCh38
rs772908130 Chr.X: 72130027-72130027 on Build GRCh38
rs746644853 Chr.X: 72130217-72130217 on Build GRCh38
rs936438936 Chr.X: 72130197-72130197 on Build GRCh38
rs767637198 Chr.X: 72130074-72130074 on Build GRCh38
rs758470258 Chr.X: 72130250-72130250 on Build GRCh38
rs745609510 Chr.X: 72130238-72130238 on Build GRCh38
rs769867866 Chr.X: 72130244-72130244 on Build GRCh38

View Full Product Details