Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5324073-5324344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR51B2
Entrez Gene ID
79345
Gene Name
olfactory receptor family 51 subfamily B member 2 (gene/pseudogene)
Gene Aliases
HOR5'Beta3, OR51B1P
UniGene
Hs.680163
Gene Chromosome Location
Chr.11: 5323298-5324352 on Build GRCh38
Transcripts
NM_033180.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508115 c.54G>T p.L18L Chr.11: 5324244-5324244 on Build GRCh38
5592272 c.72G>A p.W24* Chr.11: 5324226-5324226 on Build GRCh38
7381560 c.185C>T p.T62I Chr.11: 5324113-5324113 on Build GRCh38
6518581 c.21A>T p.A7A Chr.11: 5324277-5324277 on Build GRCh38
6518580 c.109C>T p.L37F Chr.11: 5324189-5324189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754232748 Chr.11: 5324114-5324114 on Build GRCh38
rs760965845 Chr.11: 5324110-5324110 on Build GRCh38
rs750965988 Chr.11: 5324133-5324133 on Build GRCh38
rs202240576 Chr.11: 5324316-5324316 on Build GRCh38
rs773067665 Chr.11: 5324093-5324093 on Build GRCh38
rs752851078 Chr.11: 5324105-5324105 on Build GRCh38
rs748523810 Chr.11: 5324108-5324108 on Build GRCh38
rs764397713 Chr.11: 5324201-5324201 on Build GRCh38
rs374101387 Chr.11: 5324237-5324237 on Build GRCh38
rs762948346 Chr.11: 5324095-5324095 on Build GRCh38
rs377070761 Chr.11: 5324197-5324197 on Build GRCh38
rs142767580 Chr.11: 5324176-5324176 on Build GRCh38
rs757710750 Chr.11: 5324115-5324115 on Build GRCh38
rs772053173 Chr.11: 5324272-5324272 on Build GRCh38
rs373567282 Chr.11: 5324144-5324144 on Build GRCh38
rs200263746 Chr.11: 5324286-5324286 on Build GRCh38
rs1039607416 Chr.11: 5324308-5324308 on Build GRCh38
rs923609530 Chr.11: 5324204-5324204 on Build GRCh38
rs886550864 Chr.11: 5324098-5324098 on Build GRCh38
rs759658992 Chr.11: 5324104-5324104 on Build GRCh38
rs147295690 Chr.11: 5324199-5324199 on Build GRCh38
rs749072884 Chr.11: 5324169-5324169 on Build GRCh38
rs770114575 Chr.11: 5324299-5324300 on Build GRCh38
rs530486626 Chr.11: 5324138-5324138 on Build GRCh38
rs745726815 Chr.11: 5324309-5324309 on Build GRCh38
rs774481530 Chr.11: 5324100-5324100 on Build GRCh38
rs4910750 Chr.11: 5324256-5324256 on Build GRCh38
rs199513908 Chr.11: 5324248-5324248 on Build GRCh38
rs144922727 Chr.11: 5324273-5324273 on Build GRCh38
rs749017501 Chr.11: 5324277-5324277 on Build GRCh38
rs757721243 Chr.11: 5324144-5324145 on Build GRCh38
rs375227790 Chr.11: 5324266-5324266 on Build GRCh38
rs754438636 Chr.11: 5324135-5324135 on Build GRCh38
rs769521745 Chr.11: 5324147-5324147 on Build GRCh38
rs981849409 Chr.11: 5324180-5324180 on Build GRCh38
rs771989076 Chr.11: 5324190-5324190 on Build GRCh38
rs758953693 Chr.11: 5324240-5324240 on Build GRCh38
rs755631059 Chr.11: 5324258-5324258 on Build GRCh38
rs772077485 Chr.11: 5324310-5324310 on Build GRCh38
rs765655197 Chr.11: 5324228-5324228 on Build GRCh38
rs148226586 Chr.11: 5324113-5324113 on Build GRCh38
rs574065687 Chr.11: 5324118-5324118 on Build GRCh38
rs754106494 Chr.11: 5324208-5324208 on Build GRCh38
rs990311238 Chr.11: 5324166-5324166 on Build GRCh38
rs942329369 Chr.11: 5324224-5324224 on Build GRCh38
rs554140429 Chr.11: 5324096-5324096 on Build GRCh38
rs80024156 Chr.11: 5324294-5324294 on Build GRCh38
rs770892365 Chr.11: 5324173-5324173 on Build GRCh38
rs762131875 Chr.11: 5324214-5324214 on Build GRCh38
rs746424862 Chr.11: 5324265-5324266 on Build GRCh38

View Full Product Details