Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133839383-133839656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB6B
Entrez Gene ID
51560
Gene Name
RAB6B, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.715344
Gene Chromosome Location
Chr.3: 133824235-133895847 on Build GRCh38
Transcripts
NM_016577.3 XM_011512893.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996283 c.385G>A p.D129N Chr.3: 133839522-133839522 on Build GRCh38
6529039 c.341A>G p.E114G Chr.3: 133839566-133839566 on Build GRCh38
479456 c.345G>C p.R115S Chr.3: 133839562-133839562 on Build GRCh38
3587672 c.299C>T p.S100F Chr.3: 133839608-133839608 on Build GRCh38
6011211 c.290-3C>T p.? Chr.3: 133839620-133839620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007369784 Chr.3: 133839523-133839523 on Build GRCh38
rs1045831650 Chr.3: 133839417-133839417 on Build GRCh38
rs757707005 Chr.3: 133839507-133839507 on Build GRCh38
rs750993937 Chr.3: 133839526-133839526 on Build GRCh38
rs764317220 Chr.3: 133839634-133839634 on Build GRCh38
rs774228407 Chr.3: 133839459-133839459 on Build GRCh38
rs200359535 Chr.3: 133839589-133839589 on Build GRCh38
rs775616778 Chr.3: 133839621-133839621 on Build GRCh38
rs781379933 Chr.3: 133839449-133839449 on Build GRCh38
rs1016076653 Chr.3: 133839571-133839571 on Build GRCh38
rs760929724 Chr.3: 133839482-133839482 on Build GRCh38
rs775757156 Chr.3: 133839474-133839474 on Build GRCh38
rs368174948 Chr.3: 133839557-133839557 on Build GRCh38
rs767582057 Chr.3: 133839471-133839471 on Build GRCh38
rs759662273 Chr.3: 133839481-133839496 on Build GRCh38
rs562339460 Chr.3: 133839462-133839462 on Build GRCh38
rs200086341 Chr.3: 133839480-133839480 on Build GRCh38
rs752939468 Chr.3: 133839473-133839473 on Build GRCh38
rs1031102701 Chr.3: 133839414-133839414 on Build GRCh38
rs528243699 Chr.3: 133839592-133839592 on Build GRCh38
rs542540493 Chr.3: 133839437-133839437 on Build GRCh38
rs201627683 Chr.3: 133839524-133839524 on Build GRCh38
rs997517245 Chr.3: 133839435-133839435 on Build GRCh38
rs774364285 Chr.3: 133839595-133839595 on Build GRCh38
rs543386119 Chr.3: 133839492-133839492 on Build GRCh38
rs897481838 Chr.3: 133839429-133839429 on Build GRCh38
rs573431875 Chr.3: 133839410-133839410 on Build GRCh38
rs532227495 Chr.3: 133839577-133839577 on Build GRCh38
rs41272333 Chr.3: 133839628-133839628 on Build GRCh38
rs769627212 Chr.3: 133839582-133839582 on Build GRCh38
rs772157331 Chr.3: 133839617-133839617 on Build GRCh38
rs747796727 Chr.3: 133839580-133839580 on Build GRCh38
rs754263427 Chr.3: 133839493-133839493 on Build GRCh38
rs759437158 Chr.3: 133839604-133839604 on Build GRCh38
rs201173392 Chr.3: 133839583-133839583 on Build GRCh38
rs892434456 Chr.3: 133839434-133839434 on Build GRCh38

View Full Product Details