Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 9708232-9708497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNKS
Entrez Gene ID
8658
Gene Name
tankyrase
Gene Aliases
ARTD5, PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5
UniGene
Hs.370267
Gene Chromosome Location
Chr.8: 9555229-9782346 on Build GRCh38
Transcripts
NM_003747.2 XM_006716263.3 XM_011543845.2 XM_011543846.2 XM_011543847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3413172 c.1557G>A p.P519P Chr.8: 9708471-9708471 on Build GRCh38
6805321 c.1527C>T p.L509L Chr.8: 9708441-9708441 on Build GRCh38
1458744 c.1516delA p.T508fs*33 Chr.8: 9708430-9708430 on Build GRCh38
5807532 c.1457A>G p.Y486C Chr.8: 9708371-9708371 on Build GRCh38
3651969 c.1505T>C p.L502S Chr.8: 9708419-9708419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768669480 Chr.8: 9708364-9708364 on Build GRCh38
rs767494686 Chr.8: 9708338-9708339 on Build GRCh38
rs1046509021 Chr.8: 9708398-9708398 on Build GRCh38
rs1032959021 Chr.8: 9708454-9708454 on Build GRCh38
rs756324053 Chr.8: 9708351-9708351 on Build GRCh38
rs773970709 Chr.8: 9708380-9708380 on Build GRCh38
rs763327070 Chr.8: 9708432-9708432 on Build GRCh38
rs558905291 Chr.8: 9708275-9708275 on Build GRCh38
rs762307507 Chr.8: 9708453-9708453 on Build GRCh38
rs996657468 Chr.8: 9708469-9708469 on Build GRCh38
rs188830245 Chr.8: 9708470-9708470 on Build GRCh38
rs954768462 Chr.8: 9708295-9708295 on Build GRCh38
rs79587922 Chr.8: 9708389-9708389 on Build GRCh38
rs1005745454 Chr.8: 9708307-9708307 on Build GRCh38
rs749171544 Chr.8: 9708358-9708358 on Build GRCh38
rs764681000 Chr.8: 9708441-9708441 on Build GRCh38
rs771590781 Chr.8: 9708395-9708395 on Build GRCh38
rs996265575 Chr.8: 9708292-9708292 on Build GRCh38
rs34305756 Chr.8: 9708468-9708469 on Build GRCh38
rs781607859 Chr.8: 9708332-9708332 on Build GRCh38
rs147617195 Chr.8: 9708323-9708323 on Build GRCh38
rs767843084 Chr.8: 9708457-9708457 on Build GRCh38
rs978870066 Chr.8: 9708311-9708311 on Build GRCh38
rs998790285 Chr.8: 9708442-9708442 on Build GRCh38
rs1045377176 Chr.8: 9708281-9708281 on Build GRCh38
rs903155786 Chr.8: 9708436-9708436 on Build GRCh38
rs868278108 Chr.8: 9708288-9708288 on Build GRCh38
rs372566002 Chr.8: 9708385-9708385 on Build GRCh38
rs745940550 Chr.8: 9708336-9708336 on Build GRCh38
rs375493489 Chr.8: 9708445-9708445 on Build GRCh38
rs554687334 Chr.8: 9708354-9708354 on Build GRCh38
rs772914830 Chr.8: 9708396-9708396 on Build GRCh38
rs957509089 Chr.8: 9708464-9708464 on Build GRCh38
rs1047282502 Chr.8: 9708298-9708298 on Build GRCh38
rs918890671 Chr.8: 9708282-9708282 on Build GRCh38
rs926129718 Chr.8: 9708312-9708312 on Build GRCh38
rs558892453 Chr.8: 9708273-9708273 on Build GRCh38
rs371225158 Chr.8: 9708300-9708300 on Build GRCh38
rs370231803 Chr.8: 9708471-9708471 on Build GRCh38
rs56411177 Chr.8: 9708429-9708429 on Build GRCh38
rs760164944 Chr.8: 9708406-9708406 on Build GRCh38
rs886330055 Chr.8: 9708416-9708416 on Build GRCh38
rs770365915 Chr.8: 9708411-9708411 on Build GRCh38
rs61423253 Chr.8: 9708272-9708272 on Build GRCh38
rs750316816 Chr.8: 9708339-9708339 on Build GRCh38
rs899952211 Chr.8: 9708328-9708328 on Build GRCh38
rs751842932 Chr.8: 9708450-9708450 on Build GRCh38
rs377733360 Chr.8: 9708296-9708296 on Build GRCh38

View Full Product Details