Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57308389-57308662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNKS1BP1
Entrez Gene ID
85456
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
TAB182
UniGene
Hs.530730
Gene Chromosome Location
Chr.11: 57299629-57324952 on Build GRCh38
Transcripts
NM_033396.2 XM_006718725.3 XM_011545325.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1354735 c.4104C>T p.G1368G Chr.11: 57308607-57308607 on Build GRCh38
429275 c.4053G>C p.Q1351H Chr.11: 57308658-57308658 on Build GRCh38
7236431 c.4179C>G p.P1393P Chr.11: 57308532-57308532 on Build GRCh38
1604759 c.4226C>T p.T1409I Chr.11: 57308485-57308485 on Build GRCh38
6805348 c.4120C>A p.P1374T Chr.11: 57308591-57308591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141792675 Chr.11: 57308464-57308464 on Build GRCh38
rs762092652 Chr.11: 57308607-57308607 on Build GRCh38
rs764368912 Chr.11: 57308517-57308517 on Build GRCh38
rs757290144 Chr.11: 57308449-57308449 on Build GRCh38
rs768331103 Chr.11: 57308568-57308568 on Build GRCh38
rs747765147 Chr.11: 57308572-57308572 on Build GRCh38
rs148443497 Chr.11: 57308435-57308435 on Build GRCh38
rs768243202 Chr.11: 57308482-57308482 on Build GRCh38
rs753705878 Chr.11: 57308533-57308533 on Build GRCh38
rs760785776 Chr.11: 57308595-57308595 on Build GRCh38
rs780777398 Chr.11: 57308569-57308569 on Build GRCh38
rs763267362 Chr.11: 57308498-57308498 on Build GRCh38
rs760079789 Chr.11: 57308510-57308510 on Build GRCh38
rs777723173 Chr.11: 57308446-57308446 on Build GRCh38
rs150600100 Chr.11: 57308511-57308511 on Build GRCh38
rs774758649 Chr.11: 57308504-57308504 on Build GRCh38
rs746453549 Chr.11: 57308562-57308562 on Build GRCh38
rs774760397 Chr.11: 57308582-57308582 on Build GRCh38
rs78723794 Chr.11: 57308617-57308617 on Build GRCh38
rs766732345 Chr.11: 57308414-57308414 on Build GRCh38
rs746723223 Chr.11: 57308480-57308480 on Build GRCh38
rs146653789 Chr.11: 57308583-57308583 on Build GRCh38
rs371257450 Chr.11: 57308603-57308603 on Build GRCh38
rs758343896 Chr.11: 57308555-57308555 on Build GRCh38
rs536720829 Chr.11: 57308463-57308463 on Build GRCh38
rs779930783 Chr.11: 57308634-57308634 on Build GRCh38
rs778739617 Chr.11: 57308550-57308550 on Build GRCh38
rs535959079 Chr.11: 57308428-57308428 on Build GRCh38
rs751095265 Chr.11: 57308639-57308639 on Build GRCh38
rs749024028 Chr.11: 57308448-57308448 on Build GRCh38
rs772563466 Chr.11: 57308592-57308592 on Build GRCh38
rs752766713 Chr.11: 57308512-57308512 on Build GRCh38
rs750394395 Chr.11: 57308551-57308551 on Build GRCh38
rs779209841 Chr.11: 57308613-57308614 on Build GRCh38
rs964425663 Chr.11: 57308559-57308559 on Build GRCh38
rs779447239 Chr.11: 57308557-57308557 on Build GRCh38
rs145489104 Chr.11: 57308581-57308581 on Build GRCh38
rs755058752 Chr.11: 57308423-57308423 on Build GRCh38
rs775743026 Chr.11: 57308593-57308593 on Build GRCh38
rs373622321 Chr.11: 57308542-57308542 on Build GRCh38
rs537294073 Chr.11: 57308500-57308500 on Build GRCh38
rs1032316090 Chr.11: 57308538-57308538 on Build GRCh38
rs371891458 Chr.11: 57308419-57308419 on Build GRCh38
rs1011686499 Chr.11: 57308554-57308554 on Build GRCh38
rs754154860 Chr.11: 57308606-57308606 on Build GRCh38
rs201868001 Chr.11: 57308543-57308543 on Build GRCh38
rs373278128 Chr.11: 57308422-57308422 on Build GRCh38
rs376536773 Chr.11: 57308442-57308442 on Build GRCh38
rs778665446 Chr.11: 57308456-57308456 on Build GRCh38
rs576702624 Chr.11: 57308487-57308487 on Build GRCh38
rs745797066 Chr.11: 57308458-57308458 on Build GRCh38
rs139621627 Chr.11: 57308513-57308513 on Build GRCh38
rs752924876 Chr.11: 57308436-57308436 on Build GRCh38
rs1013040936 Chr.11: 57308509-57308509 on Build GRCh38
rs758276174 Chr.11: 57308625-57308625 on Build GRCh38

View Full Product Details