Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101130224-101130495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINE1
Entrez Gene ID
5054
Gene Name
serpin family E member 1
Gene Aliases
PAI, PAI-1, PAI1, PLANH1
UniGene
Hs.414795
Gene Chromosome Location
Chr.7: 101127087-101139266 on Build GRCh38
Transcripts
NM_000602.4 XM_017012260.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1446806 c.283G>A p.A95T Chr.7: 101130432-101130432 on Build GRCh38
296954 c.312G>A p.E104E Chr.7: 101130461-101130461 on Build GRCh38
1446807 c.285C>T p.A95A Chr.7: 101130434-101130434 on Build GRCh38
6775636 c.288C>T p.P96P Chr.7: 101130437-101130437 on Build GRCh38
1446808 c.315C>A p.L105L Chr.7: 101130464-101130464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750090085 Chr.7: 101130285-101130285 on Build GRCh38
rs781571389 Chr.7: 101130445-101130445 on Build GRCh38
rs867218040 Chr.7: 101130345-101130345 on Build GRCh38
rs777658027 Chr.7: 101130437-101130437 on Build GRCh38
rs773829498 Chr.7: 101130457-101130457 on Build GRCh38
rs982863811 Chr.7: 101130352-101130352 on Build GRCh38
rs997097660 Chr.7: 101130359-101130359 on Build GRCh38
rs940008356 Chr.7: 101130430-101130431 on Build GRCh38
rs779844988 Chr.7: 101130297-101130297 on Build GRCh38
rs953312881 Chr.7: 101130369-101130369 on Build GRCh38
rs1028655368 Chr.7: 101130361-101130361 on Build GRCh38
rs756006462 Chr.7: 101130433-101130433 on Build GRCh38
rs765426708 Chr.7: 101130383-101130383 on Build GRCh38
rs1041425777 Chr.7: 101130302-101130302 on Build GRCh38
rs776711128 Chr.7: 101130403-101130404 on Build GRCh38
rs754736961 Chr.7: 101130416-101130416 on Build GRCh38
rs773381626 Chr.7: 101130398-101130398 on Build GRCh38
rs770461092 Chr.7: 101130456-101130456 on Build GRCh38
rs763181047 Chr.7: 101130461-101130461 on Build GRCh38
rs901607803 Chr.7: 101130349-101130349 on Build GRCh38
rs751805519 Chr.7: 101130415-101130415 on Build GRCh38
rs373715426 Chr.7: 101130411-101130411 on Build GRCh38
rs745457018 Chr.7: 101130442-101130442 on Build GRCh38
rs772617458 Chr.7: 101130310-101130310 on Build GRCh38
rs777007200 Chr.7: 101130374-101130374 on Build GRCh38
rs752603739 Chr.7: 101130429-101130429 on Build GRCh38
rs370282495 Chr.7: 101130311-101130311 on Build GRCh38
rs748847141 Chr.7: 101130438-101130438 on Build GRCh38
rs748629078 Chr.7: 101130453-101130453 on Build GRCh38
rs1000112229 Chr.7: 101130439-101130439 on Build GRCh38
rs762321778 Chr.7: 101130381-101130381 on Build GRCh38
rs766754145 Chr.7: 101130413-101130413 on Build GRCh38
rs144953342 Chr.7: 101130444-101130444 on Build GRCh38
rs746905446 Chr.7: 101130360-101130360 on Build GRCh38
rs893323461 Chr.7: 101130268-101130268 on Build GRCh38
rs145084838 Chr.7: 101130440-101130440 on Build GRCh38
rs773536411 Chr.7: 101130317-101130317 on Build GRCh38
rs370642281 Chr.7: 101130403-101130403 on Build GRCh38
rs969657269 Chr.7: 101130266-101130266 on Build GRCh38
rs927890081 Chr.7: 101130342-101130342 on Build GRCh38
rs767288984 Chr.7: 101130427-101130427 on Build GRCh38
rs61731296 Chr.7: 101130466-101130466 on Build GRCh38
rs908240372 Chr.7: 101130365-101130365 on Build GRCh38

View Full Product Details