Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127916293-127916548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GALNAC4
Entrez Gene ID
27090
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Gene Aliases
IV, SIAT3-C, SIAT3C, SIAT7-D, SIAT7D, ST6GALNACIV, ST6GalNAc
UniGene
Hs.3972
Gene Chromosome Location
Chr.9: 127907886-127917052 on Build GRCh38
Transcripts
NM_175039.3 NM_175040.3 XM_011518527.2 XM_011518529.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238267 c.3G>A p.M1I Chr.9: 127916417-127916417 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987544189 Chr.9: 127916358-127916358 on Build GRCh38
rs751665399 Chr.9: 127916458-127916458 on Build GRCh38
rs776395492 Chr.9: 127916421-127916421 on Build GRCh38
rs914168291 Chr.9: 127916332-127916332 on Build GRCh38
rs746820981 Chr.9: 127916397-127916397 on Build GRCh38
rs764408246 Chr.9: 127916456-127916456 on Build GRCh38
rs770412353 Chr.9: 127916375-127916375 on Build GRCh38
rs573831670 Chr.9: 127916448-127916448 on Build GRCh38
rs995439883 Chr.9: 127916521-127916521 on Build GRCh38
rs759535489 Chr.9: 127916426-127916426 on Build GRCh38
rs760102963 Chr.9: 127916372-127916373 on Build GRCh38
rs923423733 Chr.9: 127916481-127916481 on Build GRCh38
rs778635876 Chr.9: 127916376-127916376 on Build GRCh38
rs781765888 Chr.9: 127916364-127916364 on Build GRCh38
rs781763167 Chr.9: 127916403-127916403 on Build GRCh38
rs891190239 Chr.9: 127916399-127916399 on Build GRCh38
rs746321623 Chr.9: 127916369-127916369 on Build GRCh38
rs201486642 Chr.9: 127916382-127916382 on Build GRCh38
rs992309449 Chr.9: 127916326-127916326 on Build GRCh38
rs747582460 Chr.9: 127916378-127916378 on Build GRCh38
rs775596127 Chr.9: 127916432-127916432 on Build GRCh38
rs113317532 Chr.9: 127916360-127916360 on Build GRCh38
rs1027285177 Chr.9: 127916528-127916528 on Build GRCh38
rs1046738467 Chr.9: 127916488-127916488 on Build GRCh38
rs371918000 Chr.9: 127916357-127916357 on Build GRCh38
rs375089890 Chr.9: 127916493-127916493 on Build GRCh38
rs765125682 Chr.9: 127916431-127916431 on Build GRCh38
rs542566736 Chr.9: 127916487-127916487 on Build GRCh38
rs757619945 Chr.9: 127916353-127916353 on Build GRCh38
rs757655352 Chr.9: 127916462-127916462 on Build GRCh38
rs372789800 Chr.9: 127916408-127916408 on Build GRCh38
rs575704329 Chr.9: 127916520-127916520 on Build GRCh38
rs895508497 Chr.9: 127916466-127916466 on Build GRCh38
rs369549640 Chr.9: 127916365-127916365 on Build GRCh38

View Full Product Details