Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92532337-92532610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM48
Entrez Gene ID
84060
Gene Name
RNA binding motif protein 48
Gene Aliases
C7orf64, HSPC304, MGC16142
UniGene
Hs.147829
Gene Chromosome Location
Chr.7: 92528709-92540481 on Build GRCh38
Transcripts
NM_032120.3 XM_005250636.4 XM_006716149.3 XM_006716150.3 XM_017012709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6764166 c.417G>T p.K139N Chr.7: 92532518-92532518 on Build GRCh38
4895595 c.435T>C p.T145T Chr.7: 92532536-92532536 on Build GRCh38
5979397 c.395_396insA p.L135fs*13 Chr.7: 92532496-92532497 on Build GRCh38
1199019 c.315A>C p.R105S Chr.7: 92532416-92532416 on Build GRCh38
4945434 c.362A>G p.Y121C Chr.7: 92532463-92532463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031904351 Chr.7: 92532429-92532429 on Build GRCh38
rs373233331 Chr.7: 92532541-92532541 on Build GRCh38
rs754768719 Chr.7: 92532496-92532497 on Build GRCh38
rs970590836 Chr.7: 92532504-92532504 on Build GRCh38
rs377601922 Chr.7: 92532589-92532589 on Build GRCh38
rs752461175 Chr.7: 92532398-92532398 on Build GRCh38
rs369445901 Chr.7: 92532513-92532513 on Build GRCh38
rs372855385 Chr.7: 92532514-92532514 on Build GRCh38
rs997445874 Chr.7: 92532392-92532392 on Build GRCh38
rs776716667 Chr.7: 92532467-92532467 on Build GRCh38
rs778887533 Chr.7: 92532497-92532497 on Build GRCh38
rs752144367 Chr.7: 92532520-92532520 on Build GRCh38
rs376509478 Chr.7: 92532460-92532460 on Build GRCh38
rs973491469 Chr.7: 92532491-92532491 on Build GRCh38
rs956316560 Chr.7: 92532446-92532446 on Build GRCh38
rs777722916 Chr.7: 92532421-92532421 on Build GRCh38
rs750919296 Chr.7: 92532369-92532369 on Build GRCh38
rs1041147579 Chr.7: 92532572-92532572 on Build GRCh38
rs753709797 Chr.7: 92532560-92532560 on Build GRCh38
rs762156056 Chr.7: 92532495-92532495 on Build GRCh38
rs368937735 Chr.7: 92532536-92532536 on Build GRCh38
rs752232044 Chr.7: 92532523-92532523 on Build GRCh38
rs763466800 Chr.7: 92532372-92532372 on Build GRCh38
rs749173324 Chr.7: 92532440-92532440 on Build GRCh38
rs754561897 Chr.7: 92532385-92532385 on Build GRCh38
rs770089596 Chr.7: 92532503-92532503 on Build GRCh38
rs757167805 Chr.7: 92532577-92532577 on Build GRCh38
rs75703315 Chr.7: 92532521-92532521 on Build GRCh38
rs368019384 Chr.7: 92532441-92532441 on Build GRCh38
rs751049647 Chr.7: 92532380-92532380 on Build GRCh38
rs780676464 Chr.7: 92532387-92532387 on Build GRCh38
rs766846378 Chr.7: 92532516-92532516 on Build GRCh38
rs778685694 Chr.7: 92532587-92532587 on Build GRCh38
rs200740112 Chr.7: 92532557-92532557 on Build GRCh38
rs768824729 Chr.7: 92532462-92532462 on Build GRCh38
rs201482613 Chr.7: 92532534-92532534 on Build GRCh38
rs765832977 Chr.7: 92532375-92532375 on Build GRCh38
rs1021831084 Chr.7: 92532496-92532496 on Build GRCh38

View Full Product Details