Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128105526-128105799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A25
Entrez Gene ID
114789
Gene Name
solute carrier family 25 member 25
Gene Aliases
MCSC, PCSCL, SCAMC-2
UniGene
Hs.5476
Gene Chromosome Location
Chr.9: 128068200-128109258 on Build GRCh38
Transcripts
NM_001006641.3 NM_001006642.3 NM_001265614.2 NM_052901.4 NR_049766.2 XM_005251688.1 XM_005251689.4 XM_006716948.2 XM_006716949.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6182896 c.718C>T p.R240C Chr.9: 128105741-128105741 on Build GRCh38
1314487 c.770G>A p.G257E Chr.9: 128105793-128105793 on Build GRCh38
6253947 c.712G>T p.A238S Chr.9: 128105735-128105735 on Build GRCh38
6182895 c.760C>T p.R254C Chr.9: 128105741-128105741 on Build GRCh38
5766855 c.771A>T p.G257G Chr.9: 128105794-128105794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375402805 Chr.9: 128105761-128105761 on Build GRCh38
rs35862377 Chr.9: 128105699-128105700 on Build GRCh38
rs183533169 Chr.9: 128105569-128105569 on Build GRCh38
rs117481192 Chr.9: 128105646-128105646 on Build GRCh38
rs375904258 Chr.9: 128105710-128105710 on Build GRCh38
rs567855104 Chr.9: 128105623-128105623 on Build GRCh38
rs372118470 Chr.9: 128105742-128105742 on Build GRCh38
rs892170456 Chr.9: 128105570-128105570 on Build GRCh38
rs150336698 Chr.9: 128105741-128105741 on Build GRCh38
rs1007972759 Chr.9: 128105616-128105616 on Build GRCh38
rs547300865 Chr.9: 128105553-128105553 on Build GRCh38
rs1049260951 Chr.9: 128105779-128105779 on Build GRCh38
rs369170860 Chr.9: 128105698-128105698 on Build GRCh38
rs372658490 Chr.9: 128105702-128105702 on Build GRCh38
rs919518938 Chr.9: 128105757-128105757 on Build GRCh38
rs535890253 Chr.9: 128105603-128105603 on Build GRCh38
rs748301336 Chr.9: 128105777-128105777 on Build GRCh38
rs761691600 Chr.9: 128105601-128105601 on Build GRCh38
rs540885891 Chr.9: 128105681-128105681 on Build GRCh38
rs189339711 Chr.9: 128105615-128105615 on Build GRCh38
rs375622010 Chr.9: 128105693-128105693 on Build GRCh38
rs750662630 Chr.9: 128105688-128105688 on Build GRCh38
rs781070275 Chr.9: 128105749-128105749 on Build GRCh38
rs780007071 Chr.9: 128105762-128105762 on Build GRCh38
rs753817818 Chr.9: 128105760-128105760 on Build GRCh38
rs749342875 Chr.9: 128105746-128105748 on Build GRCh38
rs766359255 Chr.9: 128105754-128105754 on Build GRCh38
rs375851494 Chr.9: 128105593-128105593 on Build GRCh38
rs980619400 Chr.9: 128105564-128105564 on Build GRCh38
rs34973041 Chr.9: 128105563-128105564 on Build GRCh38
rs757894493 Chr.9: 128105723-128105723 on Build GRCh38
rs375103895 Chr.9: 128105610-128105610 on Build GRCh38
rs764825400 Chr.9: 128105713-128105713 on Build GRCh38
rs926321679 Chr.9: 128105565-128105565 on Build GRCh38
rs745836070 Chr.9: 128105753-128105753 on Build GRCh38
rs369203375 Chr.9: 128105775-128105775 on Build GRCh38
rs192337629 Chr.9: 128105671-128105671 on Build GRCh38
rs962100912 Chr.9: 128105674-128105674 on Build GRCh38
rs756328750 Chr.9: 128105689-128105690 on Build GRCh38
rs1056525389 Chr.9: 128105567-128105567 on Build GRCh38
rs116657520 Chr.9: 128105628-128105628 on Build GRCh38
rs780032348 Chr.9: 128105690-128105690 on Build GRCh38
rs185336053 Chr.9: 128105701-128105701 on Build GRCh38
rs998752275 Chr.9: 128105647-128105647 on Build GRCh38
rs946272235 Chr.9: 128105579-128105579 on Build GRCh38
rs749392034 Chr.9: 128105773-128105773 on Build GRCh38
rs769573706 Chr.9: 128105755-128105755 on Build GRCh38
rs755086874 Chr.9: 128105774-128105774 on Build GRCh38
rs939089598 Chr.9: 128105566-128105566 on Build GRCh38
rs368885931 Chr.9: 128105714-128105714 on Build GRCh38
rs774549299 Chr.9: 128105696-128105696 on Build GRCh38
rs529510298 Chr.9: 128105679-128105679 on Build GRCh38

View Full Product Details