Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 62653101-62653362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLN2
Entrez Gene ID
83660
Gene Name
talin 2
Gene Aliases
ILWEQ
UniGene
Hs.569438
Gene Chromosome Location
Chr.15: 62390527-62844631 on Build GRCh38
Transcripts
NM_015059.2 XM_005254708.4 XM_005254710.4 XM_005254711.4 XM_005254712.4 XM_005254713.4 XM_005254714.3 XM_005254715.2 XM_006720717.3 XM_017022665.1 XM_017022666.1 XM_017022667.1 XM_017022668.1 XM_017022669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585195 c.382G>C p.E128Q Chr.15: 62653179-62653179 on Build GRCh38
6802450 c.434G>A p.G145D Chr.15: 62653231-62653231 on Build GRCh38
1229386 c.373A>C p.N125H Chr.15: 62653170-62653170 on Build GRCh38
2254595 c.449A>C p.D150A Chr.15: 62653246-62653246 on Build GRCh38
1749253 c.466G>C p.D156H Chr.15: 62653263-62653263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367916448 Chr.15: 62653229-62653229 on Build GRCh38
rs1046469012 Chr.15: 62653274-62653274 on Build GRCh38
rs779713284 Chr.15: 62653341-62653341 on Build GRCh38
rs193116526 Chr.15: 62653204-62653204 on Build GRCh38
rs370300510 Chr.15: 62653182-62653182 on Build GRCh38
rs966887608 Chr.15: 62653184-62653184 on Build GRCh38
rs866879036 Chr.15: 62653263-62653263 on Build GRCh38
rs895144194 Chr.15: 62653231-62653231 on Build GRCh38
rs771777693 Chr.15: 62653194-62653194 on Build GRCh38
rs868793488 Chr.15: 62653301-62653301 on Build GRCh38
rs746122194 Chr.15: 62653214-62653214 on Build GRCh38
rs779147979 Chr.15: 62653205-62653205 on Build GRCh38
rs758653779 Chr.15: 62653339-62653339 on Build GRCh38
rs767945626 Chr.15: 62653178-62653178 on Build GRCh38
rs1030476866 Chr.15: 62653183-62653183 on Build GRCh38
rs751939066 Chr.15: 62653167-62653167 on Build GRCh38
rs776254709 Chr.15: 62653241-62653241 on Build GRCh38
rs576244868 Chr.15: 62653165-62653165 on Build GRCh38
rs142023685 Chr.15: 62653228-62653228 on Build GRCh38
rs923412471 Chr.15: 62653128-62653128 on Build GRCh38
rs929486063 Chr.15: 62653164-62653164 on Build GRCh38
rs755035467 Chr.15: 62653262-62653262 on Build GRCh38
rs778988014 Chr.15: 62653331-62653331 on Build GRCh38
rs764634075 Chr.15: 62653276-62653276 on Build GRCh38
rs761395215 Chr.15: 62653187-62653187 on Build GRCh38
rs769457345 Chr.15: 62653247-62653247 on Build GRCh38
rs753977055 Chr.15: 62653203-62653203 on Build GRCh38
rs372145437 Chr.15: 62653261-62653261 on Build GRCh38
rs772947832 Chr.15: 62653256-62653256 on Build GRCh38
rs140578416 Chr.15: 62653201-62653201 on Build GRCh38
rs761444752 Chr.15: 62653243-62653243 on Build GRCh38
rs771719157 Chr.15: 62653215-62653215 on Build GRCh38
rs925569091 Chr.15: 62653210-62653210 on Build GRCh38
rs757287982 Chr.15: 62653295-62653295 on Build GRCh38
rs1013642664 Chr.15: 62653265-62653265 on Build GRCh38
rs928448203 Chr.15: 62653334-62653334 on Build GRCh38
rs1025498369 Chr.15: 62653298-62653298 on Build GRCh38
rs745961189 Chr.15: 62653332-62653332 on Build GRCh38
rs990877752 Chr.15: 62653254-62653254 on Build GRCh38
rs573732355 Chr.15: 62653338-62653338 on Build GRCh38
rs755254947 Chr.15: 62653174-62653174 on Build GRCh38
rs768709967 Chr.15: 62653232-62653232 on Build GRCh38
rs373427269 Chr.15: 62653156-62653156 on Build GRCh38
rs753884519 Chr.15: 62653288-62653288 on Build GRCh38

View Full Product Details