Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36476734-36476959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIM1
Entrez Gene ID
51232
Gene Name
cysteine rich transmembrane BMP regulator 1
Gene Aliases
CRIM-1, S52
UniGene
Hs.699247
Gene Chromosome Location
Chr.2: 36356088-36551135 on Build GRCh38
Transcripts
NM_016441.2 XM_005264357.4 XM_011532898.2 XM_011532899.2 XM_011532901.2 XM_017004258.1 XM_017004259.1 XM_017004260.1 XM_017004261.1 XM_017004262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1020588 c.1048C>T p.R350* Chr.2: 36476945-36476945 on Build GRCh38
265524 c.1026A>T p.E342D Chr.2: 36476923-36476923 on Build GRCh38
1752536 c.1037G>C p.G346A Chr.2: 36476934-36476934 on Build GRCh38
1741732 c.1009G>A p.V337I Chr.2: 36476906-36476906 on Build GRCh38
3581355 c.1033G>A p.D345N Chr.2: 36476930-36476930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756592810 Chr.2: 36476916-36476916 on Build GRCh38
rs377405765 Chr.2: 36476906-36476906 on Build GRCh38
rs779143899 Chr.2: 36476932-36476932 on Build GRCh38
rs373942317 Chr.2: 36476875-36476875 on Build GRCh38
rs2276672 Chr.2: 36476769-36476769 on Build GRCh38
rs1041105785 Chr.2: 36476889-36476889 on Build GRCh38
rs766022040 Chr.2: 36476905-36476905 on Build GRCh38
rs768436156 Chr.2: 36476864-36476864 on Build GRCh38
rs998824069 Chr.2: 36476885-36476885 on Build GRCh38
rs754223876 Chr.2: 36476923-36476923 on Build GRCh38
rs764859631 Chr.2: 36476881-36476881 on Build GRCh38
rs775365449 Chr.2: 36476858-36476858 on Build GRCh38
rs778021611 Chr.2: 36476839-36476839 on Build GRCh38
rs915026144 Chr.2: 36476796-36476796 on Build GRCh38
rs375676368 Chr.2: 36476902-36476902 on Build GRCh38
rs746938532 Chr.2: 36476862-36476862 on Build GRCh38
rs988080552 Chr.2: 36476772-36476772 on Build GRCh38
rs776292709 Chr.2: 36476867-36476867 on Build GRCh38
rs370848114 Chr.2: 36476843-36476843 on Build GRCh38
rs561130616 Chr.2: 36476793-36476793 on Build GRCh38
rs749471444 Chr.2: 36476840-36476840 on Build GRCh38
rs771966190 Chr.2: 36476856-36476856 on Build GRCh38
rs994568892 Chr.2: 36476842-36476842 on Build GRCh38
rs34034373 Chr.2: 36476820-36476821 on Build GRCh38
rs368600447 Chr.2: 36476837-36476837 on Build GRCh38
rs778949121 Chr.2: 36476844-36476844 on Build GRCh38
rs761518996 Chr.2: 36476868-36476868 on Build GRCh38
rs966348194 Chr.2: 36476882-36476883 on Build GRCh38
rs567306685 Chr.2: 36476823-36476823 on Build GRCh38
rs757478626 Chr.2: 36476925-36476925 on Build GRCh38
rs77757898 Chr.2: 36476802-36476802 on Build GRCh38
rs1037597781 Chr.2: 36476830-36476830 on Build GRCh38
rs764454915 Chr.2: 36476918-36476918 on Build GRCh38
rs772828777 Chr.2: 36476892-36476892 on Build GRCh38
rs745995096 Chr.2: 36476847-36476847 on Build GRCh38
rs940168595 Chr.2: 36476813-36476813 on Build GRCh38

View Full Product Details