Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 33146154-33146427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS9
Entrez Gene ID
27241
Gene Name
Bardet-Biedl syndrome 9
Gene Aliases
B1, C18, D1, PTHB1
UniGene
Hs.372360
Gene Chromosome Location
Chr.7: 33129244-33637238 on Build GRCh38
Transcripts
NM_001033604.1 NM_001033605.1 NM_014451.3 NM_198428.2 XM_005249700.3 XM_005249701.2 XM_011515264.2 XM_011515265.2 XM_011515266.2 XM_011515267.2 XM_011515268.1 XM_011515269.2 XM_011515270.2 XM_017011990.1 XM_017011991.1 XM_017011992.1 XM_017011993.1 XM_017011994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752390 c.38T>G p.I13S Chr.7: 33146290-33146290 on Build GRCh38
6610187 c.19C>T p.R7C Chr.7: 33146271-33146271 on Build GRCh38
4752389 c.38T>G p.I13S Chr.7: 33146290-33146290 on Build GRCh38
6610188 c.19C>T p.R7C Chr.7: 33146271-33146271 on Build GRCh38
6610199 c.21T>C p.R7R Chr.7: 33146273-33146273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768898699 Chr.7: 33146398-33146398 on Build GRCh38
rs577536627 Chr.7: 33146357-33146357 on Build GRCh38
rs749868640 Chr.7: 33146392-33146392 on Build GRCh38
rs752284878 Chr.7: 33146369-33146369 on Build GRCh38
rs756162387 Chr.7: 33146216-33146216 on Build GRCh38
rs779638896 Chr.7: 33146310-33146310 on Build GRCh38
rs746340993 Chr.7: 33146272-33146272 on Build GRCh38
rs562061673 Chr.7: 33146250-33146250 on Build GRCh38
rs770222712 Chr.7: 33146388-33146388 on Build GRCh38
rs749116757 Chr.7: 33146244-33146244 on Build GRCh38
rs768869485 Chr.7: 33146289-33146289 on Build GRCh38
rs367892028 Chr.7: 33146380-33146380 on Build GRCh38
rs766448677 Chr.7: 33146347-33146347 on Build GRCh38
rs779974344 Chr.7: 33146223-33146223 on Build GRCh38
rs753601080 Chr.7: 33146354-33146354 on Build GRCh38
rs770310531 Chr.7: 33146279-33146279 on Build GRCh38
rs754742917 Chr.7: 33146360-33146360 on Build GRCh38
rs747632366 Chr.7: 33146270-33146270 on Build GRCh38
rs869025208 Chr.7: 33146354-33146366 on Build GRCh38
rs778823074 Chr.7: 33146362-33146362 on Build GRCh38
rs950114586 Chr.7: 33146186-33146186 on Build GRCh38
rs201415758 Chr.7: 33146339-33146339 on Build GRCh38
rs533453805 Chr.7: 33146385-33146385 on Build GRCh38
rs371653658 Chr.7: 33146383-33146383 on Build GRCh38
rs181845593 Chr.7: 33146207-33146207 on Build GRCh38
rs760638472 Chr.7: 33146340-33146340 on Build GRCh38
rs184994140 Chr.7: 33146271-33146271 on Build GRCh38
rs763234464 Chr.7: 33146288-33146288 on Build GRCh38
rs767674907 Chr.7: 33146318-33146318 on Build GRCh38
rs375087794 Chr.7: 33146206-33146206 on Build GRCh38
rs750581462 Chr.7: 33146328-33146328 on Build GRCh38
rs780556539 Chr.7: 33146389-33146389 on Build GRCh38
rs746345067 Chr.7: 33146263-33146263 on Build GRCh38
rs758084698 Chr.7: 33146370-33146370 on Build GRCh38
rs4498440 Chr.7: 33146286-33146286 on Build GRCh38
rs774422808 Chr.7: 33146307-33146307 on Build GRCh38
rs372863184 Chr.7: 33146214-33146214 on Build GRCh38
rs767764566 Chr.7: 33146213-33146213 on Build GRCh38

View Full Product Details