Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131652688-131652925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP3
Entrez Gene ID
5169
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 3
Gene Aliases
B10, CD203c, NPP3, PD-IBETA, PDNP3
UniGene
Hs.486489
Gene Chromosome Location
Chr.6: 131637235-131747413 on Build GRCh38
Transcripts
NM_005021.4 NR_133007.1 XM_011535897.1 XM_017010932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6450394 c.349C>T p.Q117* Chr.6: 131652878-131652878 on Build GRCh38
3829045 c.460G>A p.E154K Chr.6: 131652887-131652887 on Build GRCh38
6857199 c.362+2T>C p.? Chr.6: 131652893-131652893 on Build GRCh38
1073309 c.428A>G p.N143S Chr.6: 131652855-131652855 on Build GRCh38
1073310 c.461A>G p.E154G Chr.6: 131652888-131652888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770190332 Chr.6: 131652808-131652808 on Build GRCh38
rs370636173 Chr.6: 131652710-131652710 on Build GRCh38
rs1030666938 Chr.6: 131652721-131652721 on Build GRCh38
rs900325599 Chr.6: 131652708-131652708 on Build GRCh38
rs769014836 Chr.6: 131652795-131652795 on Build GRCh38
rs771790691 Chr.6: 131652890-131652890 on Build GRCh38
rs3765562 Chr.6: 131652792-131652792 on Build GRCh38
rs748021247 Chr.6: 131652888-131652888 on Build GRCh38
rs966576431 Chr.6: 131652744-131652744 on Build GRCh38
rs778826629 Chr.6: 131652708-131652708 on Build GRCh38
rs773409506 Chr.6: 131652815-131652815 on Build GRCh38
rs538375435 Chr.6: 131652786-131652786 on Build GRCh38
rs372891484 Chr.6: 131652863-131652863 on Build GRCh38
rs777009208 Chr.6: 131652805-131652805 on Build GRCh38
rs146856230 Chr.6: 131652858-131652858 on Build GRCh38
rs762992001 Chr.6: 131652821-131652821 on Build GRCh38
rs578155131 Chr.6: 131652765-131652765 on Build GRCh38
rs752585689 Chr.6: 131652844-131652844 on Build GRCh38
rs748790900 Chr.6: 131652711-131652711 on Build GRCh38
rs748631918 Chr.6: 131652807-131652807 on Build GRCh38
rs763931639 Chr.6: 131652846-131652846 on Build GRCh38
rs781663942 Chr.6: 131652893-131652893 on Build GRCh38
rs778729747 Chr.6: 131652868-131652868 on Build GRCh38
rs369646709 Chr.6: 131652887-131652887 on Build GRCh38
rs757035189 Chr.6: 131652716-131652716 on Build GRCh38
rs767270852 Chr.6: 131652842-131652842 on Build GRCh38
rs766640102 Chr.6: 131652823-131652823 on Build GRCh38
rs902572934 Chr.6: 131652800-131652800 on Build GRCh38
rs201043126 Chr.6: 131652845-131652845 on Build GRCh38
rs755312144 Chr.6: 131652886-131652886 on Build GRCh38
rs570936470 Chr.6: 131652764-131652764 on Build GRCh38
rs866507649 Chr.6: 131652717-131652717 on Build GRCh38
rs957809336 Chr.6: 131652781-131652781 on Build GRCh38
rs759361898 Chr.6: 131652838-131652838 on Build GRCh38
rs747425561 Chr.6: 131652794-131652794 on Build GRCh38
rs774510720 Chr.6: 131652833-131652833 on Build GRCh38
rs758589305 Chr.6: 131652824-131652824 on Build GRCh38
rs1055325213 Chr.6: 131652772-131652772 on Build GRCh38
rs61741318 Chr.6: 131652885-131652885 on Build GRCh38

View Full Product Details