Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41756849-41757084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM6
Entrez Gene ID
4680
Gene Name
carcinoembryonic antigen related cell adhesion molecule 6
Gene Aliases
CD66c, CEAL, NCA
UniGene
Hs.466814
Gene Chromosome Location
Chr.19: 41755421-41772211 on Build GRCh38
Transcripts
NM_002483.6 XM_011526990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1178968 c.370G>T p.V124F Chr.19: 41756905-41756905 on Build GRCh38
418223 c.324G>A p.L108L Chr.19: 41756859-41756859 on Build GRCh38
6625934 c.393T>C p.N131N Chr.19: 41756928-41756928 on Build GRCh38
439610 c.424C>T p.P142S Chr.19: 41756959-41756959 on Build GRCh38
1129828 c.385C>A p.L129I Chr.19: 41756920-41756920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111448531 Chr.19: 41757015-41757015 on Build GRCh38
rs782398702 Chr.19: 41756936-41756936 on Build GRCh38
rs782627322 Chr.19: 41756998-41756998 on Build GRCh38
rs782490833 Chr.19: 41756882-41756882 on Build GRCh38
rs782558850 Chr.19: 41756975-41756975 on Build GRCh38
rs782130617 Chr.19: 41756956-41756956 on Build GRCh38
rs1031166801 Chr.19: 41757030-41757030 on Build GRCh38
rs782773255 Chr.19: 41756960-41756960 on Build GRCh38
rs781793858 Chr.19: 41756897-41756897 on Build GRCh38
rs781867074 Chr.19: 41756879-41756879 on Build GRCh38
rs150210001 Chr.19: 41756941-41756941 on Build GRCh38
rs782302022 Chr.19: 41756900-41756900 on Build GRCh38
rs764887273 Chr.19: 41756898-41756898 on Build GRCh38
rs782788865 Chr.19: 41756873-41756873 on Build GRCh38
rs781832362 Chr.19: 41756962-41756962 on Build GRCh38
rs782700074 Chr.19: 41756968-41756968 on Build GRCh38
rs1000730833 Chr.19: 41757027-41757027 on Build GRCh38
rs116746453 Chr.19: 41756933-41756933 on Build GRCh38
rs528072396 Chr.19: 41756895-41756895 on Build GRCh38
rs113477324 Chr.19: 41757034-41757034 on Build GRCh38
rs140710603 Chr.19: 41756906-41756906 on Build GRCh38
rs143835773 Chr.19: 41756923-41756923 on Build GRCh38
rs782074909 Chr.19: 41756965-41756965 on Build GRCh38
rs548452372 Chr.19: 41756951-41756951 on Build GRCh38
rs781880546 Chr.19: 41756988-41756988 on Build GRCh38
rs919387032 Chr.19: 41756940-41756940 on Build GRCh38
rs570434430 Chr.19: 41756869-41756869 on Build GRCh38
rs531861345 Chr.19: 41756972-41756972 on Build GRCh38
rs112605825 Chr.19: 41757057-41757057 on Build GRCh38
rs782499628 Chr.19: 41756996-41756996 on Build GRCh38
rs947970697 Chr.19: 41756993-41756993 on Build GRCh38
rs903671379 Chr.19: 41757016-41757016 on Build GRCh38
rs570246433 Chr.19: 41757022-41757022 on Build GRCh38
rs1057160435 Chr.19: 41756942-41756942 on Build GRCh38
rs782673272 Chr.19: 41756987-41756987 on Build GRCh38
rs782209677 Chr.19: 41757010-41757010 on Build GRCh38
rs782619716 Chr.19: 41756885-41756885 on Build GRCh38

View Full Product Details