Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 77825357-77825584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAT1L
Entrez Gene ID
57687
Gene Name
vesicle amine transport 1-like
Gene Aliases
-
UniGene
Hs.461405
Gene Chromosome Location
Chr.16: 77788564-77980107 on Build GRCh38
Transcripts
NM_020927.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7335852 c.573_574GG>AA p.G192S Chr.16: 77825455-77825456 on Build GRCh38
1709440 c.489G>A p.M163I Chr.16: 77825371-77825371 on Build GRCh38
973926 c.502G>T p.A168S Chr.16: 77825384-77825384 on Build GRCh38
5611154 c.503C>T p.A168V Chr.16: 77825385-77825385 on Build GRCh38
4916560 c.542G>A p.G181E Chr.16: 77825424-77825424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759366220 Chr.16: 77825402-77825402 on Build GRCh38
rs529514296 Chr.16: 77825403-77825403 on Build GRCh38
rs199581723 Chr.16: 77825495-77825495 on Build GRCh38
rs777471277 Chr.16: 77825441-77825441 on Build GRCh38
rs915405653 Chr.16: 77825416-77825416 on Build GRCh38
rs1055667234 Chr.16: 77825413-77825413 on Build GRCh38
rs1028016520 Chr.16: 77825523-77825523 on Build GRCh38
rs1005147013 Chr.16: 77825547-77825547 on Build GRCh38
rs545217766 Chr.16: 77825458-77825458 on Build GRCh38
rs771690829 Chr.16: 77825440-77825440 on Build GRCh38
rs777307697 Chr.16: 77825460-77825461 on Build GRCh38
rs946916427 Chr.16: 77825418-77825418 on Build GRCh38
rs779156303 Chr.16: 77825438-77825438 on Build GRCh38
rs1048405642 Chr.16: 77825519-77825519 on Build GRCh38
rs889770347 Chr.16: 77825529-77825529 on Build GRCh38
rs766365399 Chr.16: 77825398-77825398 on Build GRCh38
rs760177189 Chr.16: 77825390-77825390 on Build GRCh38
rs746574204 Chr.16: 77825445-77825445 on Build GRCh38
rs757435504 Chr.16: 77825419-77825419 on Build GRCh38
rs765788361 Chr.16: 77825459-77825459 on Build GRCh38
rs536983813 Chr.16: 77825554-77825554 on Build GRCh38
rs144833528 Chr.16: 77825427-77825427 on Build GRCh38
rs972874282 Chr.16: 77825477-77825477 on Build GRCh38
rs377533454 Chr.16: 77825437-77825437 on Build GRCh38
rs771695160 Chr.16: 77825381-77825381 on Build GRCh38
rs762251623 Chr.16: 77825485-77825485 on Build GRCh38
rs562175352 Chr.16: 77825522-77825522 on Build GRCh38
rs918706763 Chr.16: 77825511-77825511 on Build GRCh38
rs201909513 Chr.16: 77825462-77825462 on Build GRCh38
rs1040932086 Chr.16: 77825540-77825540 on Build GRCh38
rs761855683 Chr.16: 77825451-77825451 on Build GRCh38
rs749839521 Chr.16: 77825430-77825430 on Build GRCh38
rs772599362 Chr.16: 77825388-77825388 on Build GRCh38
rs571215914 Chr.16: 77825531-77825531 on Build GRCh38
rs183892918 Chr.16: 77825399-77825399 on Build GRCh38
rs996114040 Chr.16: 77825518-77825518 on Build GRCh38
rs775155171 Chr.16: 77825463-77825463 on Build GRCh38
rs551116364 Chr.16: 77825513-77825513 on Build GRCh38
rs140225939 Chr.16: 77825461-77825461 on Build GRCh38
rs752489819 Chr.16: 77825417-77825417 on Build GRCh38
rs186873817 Chr.16: 77825428-77825428 on Build GRCh38
rs149678482 Chr.16: 77825377-77825377 on Build GRCh38
rs370193912 Chr.16: 77825412-77825412 on Build GRCh38
rs750921271 Chr.16: 77825486-77825486 on Build GRCh38
rs767773488 Chr.16: 77825420-77825420 on Build GRCh38

View Full Product Details