Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73484173-73484446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFM
Entrez Gene ID
173
Gene Name
afamin
Gene Aliases
ALB2, ALBA, ALF
UniGene
Hs.168718
Gene Chromosome Location
Chr.4: 73481740-73504001 on Build GRCh38
Transcripts
NM_001133.2 XM_017007842.1 XM_017007843.1 XM_017007844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6587039 c.166C>G p.Q56E Chr.4: 73484286-73484286 on Build GRCh38
4534803 c.211G>A p.V71I Chr.4: 73484331-73484331 on Build GRCh38
3605701 c.169G>A p.E57K Chr.4: 73484289-73484289 on Build GRCh38
1057427 c.174A>G p.A58A Chr.4: 73484294-73484294 on Build GRCh38
3605702 c.184G>A p.E62K Chr.4: 73484304-73484304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749493773 Chr.4: 73484372-73484372 on Build GRCh38
rs1014497888 Chr.4: 73484252-73484252 on Build GRCh38
rs770713150 Chr.4: 73484267-73484267 on Build GRCh38
rs780896937 Chr.4: 73484371-73484371 on Build GRCh38
rs188646494 Chr.4: 73484232-73484232 on Build GRCh38
rs764970820 Chr.4: 73484350-73484350 on Build GRCh38
rs371144053 Chr.4: 73484231-73484231 on Build GRCh38
rs367824442 Chr.4: 73484216-73484216 on Build GRCh38
rs766724290 Chr.4: 73484323-73484323 on Build GRCh38
rs977106342 Chr.4: 73484201-73484201 on Build GRCh38
rs768757281 Chr.4: 73484282-73484282 on Build GRCh38
rs756571155 Chr.4: 73484237-73484237 on Build GRCh38
rs753518215 Chr.4: 73484329-73484329 on Build GRCh38
rs746352283 Chr.4: 73484403-73484403 on Build GRCh38
rs754509549 Chr.4: 73484239-73484239 on Build GRCh38
rs372324768 Chr.4: 73484411-73484411 on Build GRCh38
rs368253144 Chr.4: 73484399-73484399 on Build GRCh38
rs780910370 Chr.4: 73484274-73484274 on Build GRCh38
rs777617182 Chr.4: 73484365-73484365 on Build GRCh38
rs1033138160 Chr.4: 73484289-73484289 on Build GRCh38
rs777707235 Chr.4: 73484260-73484260 on Build GRCh38
rs748181785 Chr.4: 73484387-73484387 on Build GRCh38
rs757913302 Chr.4: 73484358-73484358 on Build GRCh38
rs778348428 Chr.4: 73484251-73484251 on Build GRCh38
rs181593933 Chr.4: 73484383-73484383 on Build GRCh38
rs779796111 Chr.4: 73484238-73484238 on Build GRCh38
rs762016747 Chr.4: 73484291-73484291 on Build GRCh38
rs764146756 Chr.4: 73484210-73484210 on Build GRCh38
rs113909773 Chr.4: 73484352-73484604 on Build GRCh38
rs772307323 Chr.4: 73484205-73484205 on Build GRCh38
rs752091540 Chr.4: 73484211-73484211 on Build GRCh38
rs768722738 Chr.4: 73484375-73484375 on Build GRCh38
rs773870181 Chr.4: 73484299-73484299 on Build GRCh38
rs532173527 Chr.4: 73484259-73484259 on Build GRCh38
rs376128176 Chr.4: 73484255-73484255 on Build GRCh38
rs140621266 Chr.4: 73484366-73484366 on Build GRCh38
rs752096987 Chr.4: 73484352-73484352 on Build GRCh38
rs200540300 Chr.4: 73484253-73484253 on Build GRCh38
rs772282656 Chr.4: 73484391-73484391 on Build GRCh38
rs746745617 Chr.4: 73484264-73484264 on Build GRCh38
rs773855757 Chr.4: 73484392-73484392 on Build GRCh38
rs761053999 Chr.4: 73484322-73484322 on Build GRCh38
rs768090251 Chr.4: 73484292-73484292 on Build GRCh38
rs999043285 Chr.4: 73484288-73484288 on Build GRCh38
rs550289850 Chr.4: 73484279-73484279 on Build GRCh38
rs754638642 Chr.4: 73484349-73484349 on Build GRCh38
rs774332723 Chr.4: 73484283-73484283 on Build GRCh38
rs1007748618 Chr.4: 73484311-73484311 on Build GRCh38

View Full Product Details