Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102767296-102767545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX30
Entrez Gene ID
93081
Gene Name
testis expressed 30
Gene Aliases
C13orf27
UniGene
Hs.398111
Gene Chromosome Location
Chr.13: 102765888-102773821 on Build GRCh38
Transcripts
NM_001286775.1 NM_001286776.1 NM_138779.4 NR_104590.1 XM_005254097.2 XM_011521135.2 XM_011521136.2 XM_017020852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6399531 c.340G>T p.V114L Chr.13: 102767314-102767314 on Build GRCh38
1365304 c.304C>T p.L102F Chr.13: 102767350-102767350 on Build GRCh38
4880113 c.357T>C p.D119D Chr.13: 102767420-102767420 on Build GRCh38
5508635 c.337C>G p.P113A Chr.13: 102767317-102767317 on Build GRCh38
4733224 c.256C>T p.L86F Chr.13: 102767398-102767398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756293224 Chr.13: 102767497-102767497 on Build GRCh38
rs747810658 Chr.13: 102767427-102767427 on Build GRCh38
rs776013111 Chr.13: 102767448-102767448 on Build GRCh38
rs200314758 Chr.13: 102767470-102767470 on Build GRCh38
rs199524126 Chr.13: 102767404-102767404 on Build GRCh38
rs145623904 Chr.13: 102767396-102767396 on Build GRCh38
rs764481667 Chr.13: 102767353-102767353 on Build GRCh38
rs763488243 Chr.13: 102767460-102767460 on Build GRCh38
rs751908434 Chr.13: 102767391-102767391 on Build GRCh38
rs781728559 Chr.13: 102767442-102767442 on Build GRCh38
rs201320494 Chr.13: 102767379-102767379 on Build GRCh38
rs541022306 Chr.13: 102767425-102767425 on Build GRCh38
rs368810970 Chr.13: 102767347-102767348 on Build GRCh38
rs1001909083 Chr.13: 102767413-102767413 on Build GRCh38
rs768373525 Chr.13: 102767423-102767423 on Build GRCh38
rs758715652 Chr.13: 102767343-102767343 on Build GRCh38
rs765830903 Chr.13: 102767443-102767443 on Build GRCh38
rs773083061 Chr.13: 102767388-102767389 on Build GRCh38
rs201119598 Chr.13: 102767402-102767402 on Build GRCh38
rs370913802 Chr.13: 102767514-102767514 on Build GRCh38
rs780724603 Chr.13: 102767380-102767380 on Build GRCh38
rs772692993 Chr.13: 102767317-102767317 on Build GRCh38
rs764418704 Chr.13: 102767466-102767466 on Build GRCh38
rs756197219 Chr.13: 102767400-102767400 on Build GRCh38
rs750616260 Chr.13: 102767476-102767476 on Build GRCh38
rs202134897 Chr.13: 102767386-102767386 on Build GRCh38
rs754928901 Chr.13: 102767516-102767516 on Build GRCh38
rs865979576 Chr.13: 102767359-102767359 on Build GRCh38
rs751940803 Chr.13: 102767467-102767467 on Build GRCh38
rs771748684 Chr.13: 102767433-102767433 on Build GRCh38
rs749247734 Chr.13: 102767487-102767492 on Build GRCh38
rs765734836 Chr.13: 102767328-102767328 on Build GRCh38
rs760097828 Chr.13: 102767318-102767318 on Build GRCh38
rs202212224 Chr.13: 102767501-102767501 on Build GRCh38
rs749046990 Chr.13: 102767495-102767499 on Build GRCh38
rs573603152 Chr.13: 102767329-102767329 on Build GRCh38
rs767857286 Chr.13: 102767475-102767475 on Build GRCh38
rs780250522 Chr.13: 102767512-102767512 on Build GRCh38
rs781189528 Chr.13: 102767381-102767381 on Build GRCh38
rs751773294 Chr.13: 102767355-102767355 on Build GRCh38

View Full Product Details