Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135363747-135364018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT4
Entrez Gene ID
4850
Gene Name
CCR4-NOT transcription complex subunit 4
Gene Aliases
CLONE243, NOT4, NOT4H
UniGene
Hs.490224
Gene Chromosome Location
Chr.7: 135361795-135510127 on Build GRCh38
Transcripts
NM_001190847.1 NM_001190849.1 NM_001190850.1 NM_013316.3 XM_017012235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4673208 c.1748C>T p.A583V Chr.7: 135363946-135363946 on Build GRCh38
1596755 c.1730C>A p.S577Y Chr.7: 135363964-135363964 on Build GRCh38
5930267 c.1739C>T p.P580L Chr.7: 135363955-135363955 on Build GRCh38
5749425 c.1828G>A p.A610T Chr.7: 135363866-135363866 on Build GRCh38
5749426 c.1828G>A p.A610T Chr.7: 135363866-135363866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745907506 Chr.7: 135363891-135363891 on Build GRCh38
rs770951118 Chr.7: 135363929-135363929 on Build GRCh38
rs1032804283 Chr.7: 135363790-135363790 on Build GRCh38
rs746893577 Chr.7: 135363928-135363928 on Build GRCh38
rs781105581 Chr.7: 135363889-135363889 on Build GRCh38
rs957310071 Chr.7: 135363821-135363821 on Build GRCh38
rs780181408 Chr.7: 135363908-135363908 on Build GRCh38
rs752004848 Chr.7: 135363879-135363879 on Build GRCh38
rs764845164 Chr.7: 135363978-135363978 on Build GRCh38
rs759047664 Chr.7: 135363967-135363967 on Build GRCh38
rs774529944 Chr.7: 135363927-135363927 on Build GRCh38
rs966338808 Chr.7: 135363921-135363921 on Build GRCh38
rs377392481 Chr.7: 135363915-135363915 on Build GRCh38
rs972235097 Chr.7: 135363989-135363989 on Build GRCh38
rs769092265 Chr.7: 135363828-135363828 on Build GRCh38
rs776248981 Chr.7: 135363819-135363819 on Build GRCh38
rs749528170 Chr.7: 135363909-135363909 on Build GRCh38
rs751022384 Chr.7: 135363958-135363960 on Build GRCh38
rs373363789 Chr.7: 135363881-135363881 on Build GRCh38
rs759089041 Chr.7: 135363827-135363827 on Build GRCh38
rs79043969 Chr.7: 135363926-135363926 on Build GRCh38
rs758771595 Chr.7: 135363837-135363837 on Build GRCh38
rs548071392 Chr.7: 135363947-135363947 on Build GRCh38
rs765806107 Chr.7: 135363816-135363816 on Build GRCh38
rs527938251 Chr.7: 135363782-135363782 on Build GRCh38
rs11976681 Chr.7: 135363971-135363971 on Build GRCh38
rs764609745 Chr.7: 135363864-135363864 on Build GRCh38
rs756214971 Chr.7: 135363906-135363906 on Build GRCh38
rs775266666 Chr.7: 135363995-135363995 on Build GRCh38
rs771843733 Chr.7: 135363919-135363919 on Build GRCh38

View Full Product Details