Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105249347-105249578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRF1
Entrez Gene ID
2972
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Gene Aliases
BRF, BRF-1, CFDS, GTF3B, HEL-S-76p, TAF3B2, TAF3C, TAFIII90, TF3B90, TFIIIB90, hBRF
UniGene
Hs.424484
Gene Chromosome Location
Chr.14: 105209286-105315577 on Build GRCh38
Transcripts
NM_001242786.1 NM_001242787.1 NM_001242788.1 NM_001242789.1 NM_001519.3 XM_005267561.3 XM_011536672.2 XM_011536673.1 NM_033271.2 XM_005268188.3

Gene Symbol
BTBD6
Entrez Gene ID
90135
Gene Name
BTB domain containing 6
Gene Aliases
BDPL
UniGene
Hs.7367
Gene Chromosome Location
Chr.14: 105248542-105251093 on Build GRCh38
Transcripts
NM_001242786.1 NM_001242787.1 NM_001242788.1 NM_001242789.1 NM_001519.3 XM_005267561.3 XM_011536672.2 XM_011536673.1 NM_033271.2 XM_005268188.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4491942 c.163C>T p.P55S Chr.14: 105249441-105249441 on Build GRCh38
396908 c.152A>T p.E51V Chr.14: 105249430-105249430 on Build GRCh38
358906 c.117C>T p.A39A Chr.14: 105249395-105249395 on Build GRCh38
3744247 c.167A>C p.D56A Chr.14: 105249445-105249445 on Build GRCh38
6613384 c.176C>A p.P59H Chr.14: 105249454-105249454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367672571 Chr.14: 105249389-105249389 on Build GRCh38
rs768617321 Chr.14: 105249427-105249427 on Build GRCh38
rs774884347 Chr.14: 105249473-105249473 on Build GRCh38
rs367795955 Chr.14: 105249384-105249384 on Build GRCh38
rs778923848 Chr.14: 105249497-105249497 on Build GRCh38
rs771279024 Chr.14: 105249441-105249441 on Build GRCh38
rs759732279 Chr.14: 105249447-105249447 on Build GRCh38
rs959462433 Chr.14: 105249494-105249495 on Build GRCh38
rs758267376 Chr.14: 105249502-105249502 on Build GRCh38
rs750039282 Chr.14: 105249484-105249484 on Build GRCh38
rs587775213 Chr.14: 105249557-105249557 on Build GRCh38
rs769165504 Chr.14: 105249518-105249518 on Build GRCh38
rs762109382 Chr.14: 105249477-105249477 on Build GRCh38
rs749299738 Chr.14: 105249517-105249517 on Build GRCh38
rs904060023 Chr.14: 105249434-105249434 on Build GRCh38
rs202185668 Chr.14: 105249532-105249532 on Build GRCh38
rs992179381 Chr.14: 105249524-105249524 on Build GRCh38
rs371677699 Chr.14: 105249420-105249420 on Build GRCh38
rs1021200673 Chr.14: 105249400-105249400 on Build GRCh38
rs587749365 Chr.14: 105249424-105249424 on Build GRCh38
rs747048900 Chr.14: 105249411-105249411 on Build GRCh38
rs776116265 Chr.14: 105249467-105249467 on Build GRCh38
rs762232052 Chr.14: 105249530-105249530 on Build GRCh38
rs755414103 Chr.14: 105249554-105249554 on Build GRCh38
rs747907395 Chr.14: 105249437-105249437 on Build GRCh38
rs746057708 Chr.14: 105249507-105249507 on Build GRCh38
rs371498545 Chr.14: 105249505-105249505 on Build GRCh38
rs1024954954 Chr.14: 105249526-105249526 on Build GRCh38
rs764421382 Chr.14: 105249381-105249381 on Build GRCh38
rs764547810 Chr.14: 105249471-105249471 on Build GRCh38
rs755687982 Chr.14: 105249485-105249485 on Build GRCh38
rs756626813 Chr.14: 105249515-105249515 on Build GRCh38
rs770012980 Chr.14: 105249509-105249509 on Build GRCh38
rs749962417 Chr.14: 105249399-105249399 on Build GRCh38
rs751652166 Chr.14: 105249548-105249548 on Build GRCh38
rs201575024 Chr.14: 105249490-105249490 on Build GRCh38
rs780773995 Chr.14: 105249483-105249483 on Build GRCh38
rs377166086 Chr.14: 105249456-105249456 on Build GRCh38
rs587671195 Chr.14: 105249386-105249386 on Build GRCh38
rs181120393 Chr.14: 105249544-105249544 on Build GRCh38
rs763475014 Chr.14: 105249470-105249470 on Build GRCh38
rs767258177 Chr.14: 105249481-105249481 on Build GRCh38
rs587700740 Chr.14: 105249442-105249442 on Build GRCh38
rs587616624 Chr.14: 105249432-105249432 on Build GRCh38
rs747967041 Chr.14: 105249500-105249500 on Build GRCh38
rs769158617 Chr.14: 105249524-105249524 on Build GRCh38
rs774825295 Chr.14: 105249525-105249525 on Build GRCh38
rs747453678 Chr.14: 105249490-105249503 on Build GRCh38
rs753336338 Chr.14: 105249493-105249493 on Build GRCh38
rs370475637 Chr.14: 105249482-105249482 on Build GRCh38
rs755566101 Chr.14: 105249410-105249410 on Build GRCh38

View Full Product Details