Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17823244-17823469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL8
Entrez Gene ID
81569
Gene Name
actin like 8
Gene Aliases
CT57
UniGene
Hs.2149
Gene Chromosome Location
Chr.1: 17755313-17827063 on Build GRCh38
Transcripts
NM_030812.2 XM_011542212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3803075 c.241C>A p.L81I Chr.1: 17823249-17823249 on Build GRCh38
5433001 c.297C>A p.I99I Chr.1: 17823305-17823305 on Build GRCh38
383704 c.280G>C p.E94Q Chr.1: 17823288-17823288 on Build GRCh38
1290059 c.302C>T p.T101M Chr.1: 17823310-17823310 on Build GRCh38
7085916 c.326C>T p.A109V Chr.1: 17823334-17823334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771216444 Chr.1: 17823360-17823360 on Build GRCh38
rs930677861 Chr.1: 17823434-17823434 on Build GRCh38
rs114174981 Chr.1: 17823385-17823385 on Build GRCh38
rs377204196 Chr.1: 17823323-17823323 on Build GRCh38
rs553710705 Chr.1: 17823373-17823373 on Build GRCh38
rs765307312 Chr.1: 17823335-17823335 on Build GRCh38
rs371861959 Chr.1: 17823292-17823292 on Build GRCh38
rs758151936 Chr.1: 17823392-17823392 on Build GRCh38
rs752015023 Chr.1: 17823280-17823280 on Build GRCh38
rs773447718 Chr.1: 17823359-17823359 on Build GRCh38
rs779078248 Chr.1: 17823278-17823279 on Build GRCh38
rs199677202 Chr.1: 17823311-17823311 on Build GRCh38
rs751469963 Chr.1: 17823408-17823408 on Build GRCh38
rs115398046 Chr.1: 17823297-17823297 on Build GRCh38
rs1036281377 Chr.1: 17823369-17823369 on Build GRCh38
rs758261864 Chr.1: 17823312-17823312 on Build GRCh38
rs200147694 Chr.1: 17823310-17823310 on Build GRCh38
rs867140619 Chr.1: 17823436-17823436 on Build GRCh38
rs200788751 Chr.1: 17823366-17823366 on Build GRCh38
rs761441930 Chr.1: 17823379-17823379 on Build GRCh38
rs746930448 Chr.1: 17823332-17823332 on Build GRCh38
rs115824043 Chr.1: 17823340-17823340 on Build GRCh38
rs759993038 Chr.1: 17823368-17823368 on Build GRCh38
rs768803261 Chr.1: 17823334-17823334 on Build GRCh38
rs753394597 Chr.1: 17823378-17823378 on Build GRCh38
rs779732117 Chr.1: 17823402-17823402 on Build GRCh38
rs199647836 Chr.1: 17823391-17823391 on Build GRCh38
rs920847630 Chr.1: 17823397-17823397 on Build GRCh38
rs770203610 Chr.1: 17823349-17823349 on Build GRCh38
rs150792142 Chr.1: 17823279-17823279 on Build GRCh38
rs760175883 Chr.1: 17823295-17823295 on Build GRCh38
rs571027502 Chr.1: 17823308-17823308 on Build GRCh38

View Full Product Details