Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 129746141-129746412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPNPEP2
Entrez Gene ID
7512
Gene Name
X-prolyl aminopeptidase 2
Gene Aliases
AEACEI, APP2
UniGene
Hs.170499
Gene Chromosome Location
Chr.X: 129738970-129769549 on Build GRCh38
Transcripts
NM_003399.5

SNP Detail

SNP ID Chromosome Location
rs768627577 Chr.X: 129746374-129746374 on Build GRCh38
rs747077411 Chr.X: 129746351-129746351 on Build GRCh38
rs769016610 Chr.X: 129746284-129746284 on Build GRCh38
rs775302431 Chr.X: 129746249-129746249 on Build GRCh38
rs750944885 Chr.X: 129746302-129746302 on Build GRCh38
rs779096426 Chr.X: 129746335-129746335 on Build GRCh38
rs745842830 Chr.X: 129746239-129746239 on Build GRCh38
rs779116485 Chr.X: 129746210-129746210 on Build GRCh38
rs774117252 Chr.X: 129746376-129746376 on Build GRCh38
rs386352381 Chr.X: 129746336-129746336 on Build GRCh38
rs749637935 Chr.X: 129746197-129746197 on Build GRCh38
rs866955033 Chr.X: 129746266-129746266 on Build GRCh38
rs749052932 Chr.X: 129746373-129746373 on Build GRCh38
rs935586572 Chr.X: 129746268-129746268 on Build GRCh38
rs756404758 Chr.X: 129746188-129746188 on Build GRCh38
rs755316061 Chr.X: 129746325-129746325 on Build GRCh38
rs367980077 Chr.X: 129746276-129746276 on Build GRCh38
rs200638074 Chr.X: 129746313-129746313 on Build GRCh38
rs780517400 Chr.X: 129746189-129746189 on Build GRCh38
rs774159719 Chr.X: 129746277-129746277 on Build GRCh38
rs763735445 Chr.X: 129746272-129746272 on Build GRCh38
rs748170691 Chr.X: 129746212-129746212 on Build GRCh38
rs181163075 Chr.X: 129746347-129746347 on Build GRCh38
rs779858853 Chr.X: 129746354-129746354 on Build GRCh38
rs41301505 Chr.X: 129746164-129746164 on Build GRCh38
rs752986980 Chr.X: 129746344-129746344 on Build GRCh38
rs200904947 Chr.X: 129746305-129746305 on Build GRCh38
rs769731066 Chr.X: 129746352-129746352 on Build GRCh38
rs762687931 Chr.X: 129746254-129746254 on Build GRCh38
rs767178210 Chr.X: 129746300-129746300 on Build GRCh38
rs769450438 Chr.X: 129746247-129746247 on Build GRCh38
rs755192507 Chr.X: 129746204-129746204 on Build GRCh38
rs761195997 Chr.X: 129746304-129746304 on Build GRCh38
rs772201096 Chr.X: 129746222-129746222 on Build GRCh38
rs140165401 Chr.X: 129746230-129746230 on Build GRCh38
rs1038830806 Chr.X: 129746323-129746323 on Build GRCh38
rs141017709 Chr.X: 129746310-129746310 on Build GRCh38
rs151094684 Chr.X: 129746236-129746236 on Build GRCh38

View Full Product Details