Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7709112-7709343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB3
Entrez Gene ID
1949
Gene Name
ephrin B3
Gene Aliases
EFL6, EPLG8, LERK8
UniGene
Hs.26988
Gene Chromosome Location
Chr.17: 7705202-7711375 on Build GRCh38
Transcripts
NM_001406.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418432 c.745A>T p.M249L Chr.17: 7709298-7709298 on Build GRCh38
5853620 c.731G>A p.G244E Chr.17: 7709284-7709284 on Build GRCh38
707842 c.709C>T p.L237L Chr.17: 7709262-7709262 on Build GRCh38
5933563 c.664C>T p.P222S Chr.17: 7709217-7709217 on Build GRCh38
5988605 c.698G>T p.G233V Chr.17: 7709251-7709251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763073256 Chr.17: 7709286-7709286 on Build GRCh38
rs1047046842 Chr.17: 7709164-7709164 on Build GRCh38
rs748865873 Chr.17: 7709258-7709258 on Build GRCh38
rs755693925 Chr.17: 7709314-7709314 on Build GRCh38
rs747602208 Chr.17: 7709248-7709248 on Build GRCh38
rs959088807 Chr.17: 7709229-7709229 on Build GRCh38
rs751856372 Chr.17: 7709307-7709307 on Build GRCh38
rs758749640 Chr.17: 7709208-7709208 on Build GRCh38
rs375397163 Chr.17: 7709308-7709308 on Build GRCh38
rs752118716 Chr.17: 7709218-7709218 on Build GRCh38
rs767965285 Chr.17: 7709311-7709311 on Build GRCh38
rs761944344 Chr.17: 7709170-7709170 on Build GRCh38
rs968731035 Chr.17: 7709252-7709252 on Build GRCh38
rs769356691 Chr.17: 7709251-7709251 on Build GRCh38
rs752145569 Chr.17: 7709249-7709250 on Build GRCh38
rs765440747 Chr.17: 7709177-7709177 on Build GRCh38
rs190525302 Chr.17: 7709152-7709152 on Build GRCh38
rs1000399298 Chr.17: 7709213-7709213 on Build GRCh38
rs770804331 Chr.17: 7709259-7709259 on Build GRCh38
rs1002710201 Chr.17: 7709185-7709185 on Build GRCh38
rs750774945 Chr.17: 7709193-7709193 on Build GRCh38
rs761902972 Chr.17: 7709271-7709271 on Build GRCh38
rs886907023 Chr.17: 7709172-7709172 on Build GRCh38
rs776862628 Chr.17: 7709270-7709270 on Build GRCh38
rs759519825 Chr.17: 7709137-7709137 on Build GRCh38
rs749024142 Chr.17: 7709321-7709321 on Build GRCh38
rs954849989 Chr.17: 7709298-7709298 on Build GRCh38
rs774475041 Chr.17: 7709133-7709133 on Build GRCh38
rs774270981 Chr.17: 7709260-7709260 on Build GRCh38
rs755473664 Chr.17: 7709237-7709237 on Build GRCh38
rs553677598 Chr.17: 7709217-7709217 on Build GRCh38
rs777129812 Chr.17: 7709320-7709320 on Build GRCh38
rs752182878 Chr.17: 7709313-7709313 on Build GRCh38
rs903558637 Chr.17: 7709194-7709194 on Build GRCh38
rs765408246 Chr.17: 7709276-7709276 on Build GRCh38
rs766797412 Chr.17: 7709291-7709291 on Build GRCh38
rs572387959 Chr.17: 7709261-7709261 on Build GRCh38
rs1036161331 Chr.17: 7709192-7709192 on Build GRCh38
rs780687356 Chr.17: 7709245-7709245 on Build GRCh38
rs750698681 Chr.17: 7709277-7709277 on Build GRCh38
rs562025360 Chr.17: 7709137-7709138 on Build GRCh38
rs866185868 Chr.17: 7709294-7709294 on Build GRCh38

View Full Product Details