Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143043220-143043457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf31
Entrez Gene ID
286122
Gene Name
chromosome 8 open reading frame 31
Gene Aliases
-
UniGene
Hs.660382
Gene Chromosome Location
Chr.8: 143038983-143054303 on Build GRCh38
Transcripts
NM_173687.3 NR_126398.1 NR_126399.1 XM_017013344.1 XM_017013345.1 XM_017013346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4668027 c.179C>T p.T60M Chr.8: 143043255-143043255 on Build GRCh38
79588 c.150C>G p.P50P Chr.8: 143043226-143043226 on Build GRCh38
6441598 c.149C>T p.P50L Chr.8: 143043225-143043225 on Build GRCh38
256506 c.146C>A p.S49Y Chr.8: 143043222-143043222 on Build GRCh38
5811570 c.158A>T p.Q53L Chr.8: 143043234-143043234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574803305 Chr.8: 143043241-143043241 on Build GRCh38
rs371751268 Chr.8: 143043350-143043350 on Build GRCh38
rs780507123 Chr.8: 143043276-143043276 on Build GRCh38
rs182304372 Chr.8: 143043384-143043384 on Build GRCh38
rs750042198 Chr.8: 143043263-143043263 on Build GRCh38
rs34474166 Chr.8: 143043385-143043385 on Build GRCh38
rs117102312 Chr.8: 143043288-143043288 on Build GRCh38
rs942716355 Chr.8: 143043382-143043382 on Build GRCh38
rs954111868 Chr.8: 143043380-143043380 on Build GRCh38
rs372593208 Chr.8: 143043255-143043255 on Build GRCh38
rs917436282 Chr.8: 143043280-143043280 on Build GRCh38
rs377256241 Chr.8: 143043285-143043285 on Build GRCh38
rs559323080 Chr.8: 143043393-143043393 on Build GRCh38
rs755610420 Chr.8: 143043281-143043281 on Build GRCh38
rs748253234 Chr.8: 143043286-143043286 on Build GRCh38
rs973420789 Chr.8: 143043426-143043426 on Build GRCh38
rs760740236 Chr.8: 143043296-143043296 on Build GRCh38
rs751132290 Chr.8: 143043256-143043256 on Build GRCh38
rs993408928 Chr.8: 143043338-143043338 on Build GRCh38
rs756357815 Chr.8: 143043247-143043247 on Build GRCh38
rs749594431 Chr.8: 143043277-143043277 on Build GRCh38
rs1009911201 Chr.8: 143043335-143043335 on Build GRCh38
rs756464085 Chr.8: 143043271-143043271 on Build GRCh38
rs979791521 Chr.8: 143043313-143043313 on Build GRCh38
rs901260425 Chr.8: 143043397-143043397 on Build GRCh38
rs1053388209 Chr.8: 143043315-143043315 on Build GRCh38
rs747299539 Chr.8: 143043305-143043305 on Build GRCh38
rs762930633 Chr.8: 143043244-143043244 on Build GRCh38
rs377481499 Chr.8: 143043411-143043411 on Build GRCh38
rs543256008 Chr.8: 143043379-143043379 on Build GRCh38
rs950204730 Chr.8: 143043282-143043282 on Build GRCh38
rs76199662 Chr.8: 143043377-143043377 on Build GRCh38
rs369570839 Chr.8: 143043342-143043342 on Build GRCh38
rs773339511 Chr.8: 143043304-143043304 on Build GRCh38
rs961386430 Chr.8: 143043403-143043403 on Build GRCh38
rs767167324 Chr.8: 143043259-143043259 on Build GRCh38
rs759608885 Chr.8: 143043396-143043396 on Build GRCh38
rs763143489 Chr.8: 143043319-143043319 on Build GRCh38

View Full Product Details