Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 19182563-19182836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRP3
Entrez Gene ID
8048
Gene Name
cysteine and glycine rich protein 3
Gene Aliases
CLP, CMD1M, CMH12, CRP3, LMO4, MLP
UniGene
Hs.83577
Gene Chromosome Location
Chr.11: 19182030-19210571 on Build GRCh38
Transcripts
NM_003476.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928943 c.571G>T p.E191* Chr.11: 19182684-19182684 on Build GRCh38
5999961 c.538G>A p.G180S Chr.11: 19182717-19182717 on Build GRCh38
304411 c.559A>G p.T187A Chr.11: 19182696-19182696 on Build GRCh38
5372225 c.576G>A p.K192K Chr.11: 19182679-19182679 on Build GRCh38
5913166 c.580G>A p.E194K Chr.11: 19182675-19182675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758986854 Chr.11: 19182786-19182786 on Build GRCh38
rs773875522 Chr.11: 19182782-19182782 on Build GRCh38
rs534055346 Chr.11: 19182642-19182643 on Build GRCh38
rs773051385 Chr.11: 19182776-19182776 on Build GRCh38
rs539346380 Chr.11: 19182779-19182779 on Build GRCh38
rs779471970 Chr.11: 19182641-19182641 on Build GRCh38
rs995015970 Chr.11: 19182724-19182724 on Build GRCh38
rs770760020 Chr.11: 19182744-19182744 on Build GRCh38
rs727504575 Chr.11: 19182739-19182739 on Build GRCh38
rs397516858 Chr.11: 19182748-19182749 on Build GRCh38
rs372220068 Chr.11: 19182671-19182671 on Build GRCh38
rs189477598 Chr.11: 19182663-19182663 on Build GRCh38
rs886038811 Chr.11: 19182680-19182680 on Build GRCh38
rs765066102 Chr.11: 19182700-19182700 on Build GRCh38
rs749611828 Chr.11: 19182796-19182797 on Build GRCh38
rs397516859 Chr.11: 19182720-19182720 on Build GRCh38
rs141100524 Chr.11: 19182679-19182679 on Build GRCh38
rs45607943 Chr.11: 19182658-19182658 on Build GRCh38
rs370147131 Chr.11: 19182731-19182731 on Build GRCh38
rs145073736 Chr.11: 19182687-19182687 on Build GRCh38
rs776527888 Chr.11: 19182769-19182769 on Build GRCh38
rs111868331 Chr.11: 19182711-19182711 on Build GRCh38
rs571722496 Chr.11: 19182675-19182675 on Build GRCh38
rs936525460 Chr.11: 19182603-19182603 on Build GRCh38
rs111525588 Chr.11: 19182655-19182655 on Build GRCh38
rs531722178 Chr.11: 19182661-19182661 on Build GRCh38
rs776339605 Chr.11: 19182690-19182690 on Build GRCh38
rs886048096 Chr.11: 19182718-19182718 on Build GRCh38
rs142019584 Chr.11: 19182719-19182719 on Build GRCh38
rs963247787 Chr.11: 19182591-19182591 on Build GRCh38
rs769573413 Chr.11: 19182773-19182773 on Build GRCh38
rs565668237 Chr.11: 19182770-19182770 on Build GRCh38
rs886038810 Chr.11: 19182683-19182683 on Build GRCh38
rs767944340 Chr.11: 19182722-19182722 on Build GRCh38
rs752103761 Chr.11: 19182644-19182644 on Build GRCh38
rs943580857 Chr.11: 19182704-19182704 on Build GRCh38
rs778319057 Chr.11: 19182726-19182726 on Build GRCh38
rs181530945 Chr.11: 19182694-19182694 on Build GRCh38
rs756715438 Chr.11: 19182739-19182739 on Build GRCh38
rs757882490 Chr.11: 19182640-19182640 on Build GRCh38
rs755498404 Chr.11: 19182645-19182645 on Build GRCh38
rs975698920 Chr.11: 19182609-19182609 on Build GRCh38
rs904202473 Chr.11: 19182714-19182714 on Build GRCh38
rs774359383 Chr.11: 19182672-19182672 on Build GRCh38
rs376799144 Chr.11: 19182742-19182742 on Build GRCh38
rs727502946 Chr.11: 19182673-19182676 on Build GRCh38
rs868344115 Chr.11: 19182681-19182681 on Build GRCh38
rs748531257 Chr.11: 19182659-19182659 on Build GRCh38
rs749887485 Chr.11: 19182625-19182625 on Build GRCh38
rs727504899 Chr.11: 19182716-19182716 on Build GRCh38
rs368444918 Chr.11: 19182662-19182662 on Build GRCh38
rs371700321 Chr.11: 19182762-19182762 on Build GRCh38
rs749515242 Chr.11: 19182736-19182736 on Build GRCh38
rs547374738 Chr.11: 19182717-19182717 on Build GRCh38
rs755450152 Chr.11: 19182721-19182721 on Build GRCh38
rs768908840 Chr.11: 19182797-19182797 on Build GRCh38
rs749568462 Chr.11: 19182664-19182664 on Build GRCh38
rs375020550 Chr.11: 19182766-19182766 on Build GRCh38
rs749002955 Chr.11: 19182723-19182723 on Build GRCh38
rs113174709 Chr.11: 19182777-19182777 on Build GRCh38

View Full Product Details