Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109102678-109102951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNG
Entrez Gene ID
7374
Gene Name
uracil DNA glycosylase
Gene Aliases
DGU, HIGM4, HIGM5, UDG, UNG1, UNG15, UNG2
UniGene
Hs.191334
Gene Chromosome Location
Chr.12: 109097594-109110993 on Build GRCh38
Transcripts
NM_003362.3 NM_080911.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
934684 c.556C>T p.H186Y Chr.12: 109102888-109102888 on Build GRCh38
4554120 c.584G>A p.W195* Chr.12: 109102916-109102916 on Build GRCh38
3376105 c.560C>A p.S187Y Chr.12: 109102865-109102865 on Build GRCh38
934682 c.536C>T p.T179I Chr.12: 109102868-109102868 on Build GRCh38
1746722 c.572A>T p.D191V Chr.12: 109102904-109102904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201388456 Chr.12: 109102898-109102898 on Build GRCh38
rs200073522 Chr.12: 109102815-109102815 on Build GRCh38
rs3219233 Chr.12: 109102750-109102750 on Build GRCh38
rs772060123 Chr.12: 109102865-109102865 on Build GRCh38
rs199547586 Chr.12: 109102884-109102884 on Build GRCh38
rs745764048 Chr.12: 109102853-109102853 on Build GRCh38
rs1037677500 Chr.12: 109102748-109102748 on Build GRCh38
rs986389540 Chr.12: 109102752-109102752 on Build GRCh38
rs772764942 Chr.12: 109102873-109102874 on Build GRCh38
rs397825341 Chr.12: 109102814-109102815 on Build GRCh38
rs398048308 Chr.12: 109102815-109102816 on Build GRCh38
rs1020427742 Chr.12: 109102826-109102826 on Build GRCh38
rs1010413399 Chr.12: 109102825-109102825 on Build GRCh38
rs937956274 Chr.12: 109102906-109102906 on Build GRCh38
rs747306480 Chr.12: 109102849-109102850 on Build GRCh38
rs769977037 Chr.12: 109102875-109102875 on Build GRCh38
rs989581223 Chr.12: 109102794-109102794 on Build GRCh38
rs769537890 Chr.12: 109102855-109102856 on Build GRCh38
rs753458087 Chr.12: 109102831-109102831 on Build GRCh38
rs958220287 Chr.12: 109102773-109102773 on Build GRCh38
rs3219235 Chr.12: 109102808-109102809 on Build GRCh38
rs941257276 Chr.12: 109102703-109102703 on Build GRCh38
rs767932993 Chr.12: 109102818-109102818 on Build GRCh38
rs758287869 Chr.12: 109102705-109102705 on Build GRCh38
rs780464784 Chr.12: 109102811-109102815 on Build GRCh38
rs756752692 Chr.12: 109102832-109102832 on Build GRCh38
rs370142146 Chr.12: 109102796-109102796 on Build GRCh38
rs777618710 Chr.12: 109102822-109102822 on Build GRCh38
rs779824297 Chr.12: 109102868-109102868 on Build GRCh38
rs753169678 Chr.12: 109102821-109102821 on Build GRCh38
rs397729718 Chr.12: 109102813-109102814 on Build GRCh38
rs182850249 Chr.12: 109102722-109102722 on Build GRCh38
rs1057213743 Chr.12: 109102776-109102776 on Build GRCh38
rs374467296 Chr.12: 109102854-109102854 on Build GRCh38
rs544644925 Chr.12: 109102730-109102730 on Build GRCh38
rs749500595 Chr.12: 109102885-109102885 on Build GRCh38
rs3219234 Chr.12: 109102763-109102763 on Build GRCh38
rs746843730 Chr.12: 109102873-109102873 on Build GRCh38
rs201032593 Chr.12: 109102816-109102816 on Build GRCh38
rs752062644 Chr.12: 109102791-109102791 on Build GRCh38
rs545691651 Chr.12: 109102849-109102849 on Build GRCh38

View Full Product Details