Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143977803-143978052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP10
Entrez Gene ID
84875
Gene Name
poly(ADP-ribose) polymerase family member 10
Gene Aliases
ARTD10
UniGene
Hs.348609
Gene Chromosome Location
Chr.8: 143977152-143986471 on Build GRCh38
Transcripts
NM_001317895.1 NM_032789.4 NR_134234.1 XM_011517336.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014830 c.2711G>A p.R904H Chr.8: 143977927-143977927 on Build GRCh38
5710882 c.2663G>C p.G888A Chr.8: 143977975-143977975 on Build GRCh38
4711165 c.2726G>A p.R909H Chr.8: 143977912-143977912 on Build GRCh38
6737817 c.2675C>T p.P892L Chr.8: 143977963-143977963 on Build GRCh38
4404313 c.2729A>G p.N910S Chr.8: 143977909-143977909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782188047 Chr.8: 143977846-143977846 on Build GRCh38
rs782620268 Chr.8: 143977913-143977913 on Build GRCh38
rs202087846 Chr.8: 143977871-143977871 on Build GRCh38
rs781949313 Chr.8: 143977853-143977853 on Build GRCh38
rs782357845 Chr.8: 143977953-143977953 on Build GRCh38
rs781847703 Chr.8: 143977971-143977971 on Build GRCh38
rs372978210 Chr.8: 143977928-143977928 on Build GRCh38
rs782809190 Chr.8: 143977891-143977891 on Build GRCh38
rs782183194 Chr.8: 143977942-143977942 on Build GRCh38
rs147608960 Chr.8: 143977844-143977844 on Build GRCh38
rs782714452 Chr.8: 143977865-143977865 on Build GRCh38
rs782213808 Chr.8: 143978009-143978009 on Build GRCh38
rs781886694 Chr.8: 143977912-143977912 on Build GRCh38
rs370678470 Chr.8: 143978006-143978006 on Build GRCh38
rs370011427 Chr.8: 143977904-143977904 on Build GRCh38
rs782366382 Chr.8: 143977848-143977848 on Build GRCh38
rs782686487 Chr.8: 143978014-143978014 on Build GRCh38
rs377690585 Chr.8: 143977893-143977893 on Build GRCh38
rs782449748 Chr.8: 143978000-143978000 on Build GRCh38
rs781854102 Chr.8: 143977987-143977987 on Build GRCh38
rs782366827 Chr.8: 143977877-143977878 on Build GRCh38
rs1050768240 Chr.8: 143977995-143977995 on Build GRCh38
rs374424555 Chr.8: 143977883-143977883 on Build GRCh38
rs782543074 Chr.8: 143977828-143977828 on Build GRCh38
rs782098765 Chr.8: 143977872-143977872 on Build GRCh38
rs916822364 Chr.8: 143977929-143977929 on Build GRCh38
rs782282328 Chr.8: 143977917-143977917 on Build GRCh38
rs782090223 Chr.8: 143977976-143977976 on Build GRCh38
rs902164096 Chr.8: 143977985-143977985 on Build GRCh38
rs376288904 Chr.8: 143977956-143977956 on Build GRCh38
rs782698203 Chr.8: 143977969-143977969 on Build GRCh38
rs782624467 Chr.8: 143977903-143977903 on Build GRCh38
rs782544946 Chr.8: 143977989-143977989 on Build GRCh38
rs201196278 Chr.8: 143977922-143977922 on Build GRCh38
rs935086047 Chr.8: 143977990-143977990 on Build GRCh38
rs782547004 Chr.8: 143977963-143977963 on Build GRCh38
rs374828498 Chr.8: 143977859-143977859 on Build GRCh38
rs533456425 Chr.8: 143977968-143977968 on Build GRCh38
rs782122072 Chr.8: 143977960-143977960 on Build GRCh38
rs983495172 Chr.8: 143977890-143977890 on Build GRCh38
rs782444673 Chr.8: 143977899-143977899 on Build GRCh38
rs182781977 Chr.8: 143977857-143977857 on Build GRCh38
rs782780930 Chr.8: 143977874-143977874 on Build GRCh38
rs782668875 Chr.8: 143977997-143977997 on Build GRCh38
rs140539975 Chr.8: 143977999-143977999 on Build GRCh38
rs374135187 Chr.8: 143978026-143978026 on Build GRCh38
rs782378617 Chr.8: 143978010-143978010 on Build GRCh38
rs782774080 Chr.8: 143977983-143977983 on Build GRCh38

View Full Product Details