Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35813646-35813879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT2
Entrez Gene ID
84681
Gene Name
histidine triad nucleotide binding protein 2
Gene Aliases
HIT-17
UniGene
Hs.70573
Gene Chromosome Location
Chr.9: 35812960-35815045 on Build GRCh38
Transcripts
NM_032593.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4626454 c.132G>A p.Q44Q Chr.9: 35813734-35813734 on Build GRCh38
244903 c.212A>G p.E71G Chr.9: 35813654-35813654 on Build GRCh38
216843 c.180G>A p.L60L Chr.9: 35813686-35813686 on Build GRCh38
216842 c.181G>A p.D61N Chr.9: 35813685-35813685 on Build GRCh38
6679948 c.179T>G p.L60R Chr.9: 35813687-35813687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756601589 Chr.9: 35813683-35813683 on Build GRCh38
rs781538171 Chr.9: 35813800-35813800 on Build GRCh38
rs748522915 Chr.9: 35813707-35813707 on Build GRCh38
rs778452052 Chr.9: 35813833-35813833 on Build GRCh38
rs747136493 Chr.9: 35813693-35813693 on Build GRCh38
rs368191656 Chr.9: 35813797-35813797 on Build GRCh38
rs778817978 Chr.9: 35813671-35813671 on Build GRCh38
rs770230594 Chr.9: 35813823-35813823 on Build GRCh38
rs748568033 Chr.9: 35813807-35813807 on Build GRCh38
rs753700264 Chr.9: 35813769-35813769 on Build GRCh38
rs758361876 Chr.9: 35813686-35813686 on Build GRCh38
rs759104993 Chr.9: 35813727-35813727 on Build GRCh38
rs779942354 Chr.9: 35813794-35813794 on Build GRCh38
rs775210200 Chr.9: 35813757-35813757 on Build GRCh38
rs374552485 Chr.9: 35813768-35813768 on Build GRCh38
rs773829371 Chr.9: 35813724-35813724 on Build GRCh38
rs771441217 Chr.9: 35813745-35813745 on Build GRCh38
rs780355885 Chr.9: 35813687-35813687 on Build GRCh38
rs761573672 Chr.9: 35813780-35813780 on Build GRCh38
rs770467624 Chr.9: 35813715-35813715 on Build GRCh38
rs572961753 Chr.9: 35813781-35813781 on Build GRCh38
rs763311269 Chr.9: 35813697-35813699 on Build GRCh38
rs973536577 Chr.9: 35813840-35813840 on Build GRCh38
rs998094333 Chr.9: 35813712-35813718 on Build GRCh38
rs758511936 Chr.9: 35813792-35813792 on Build GRCh38
rs574817962 Chr.9: 35813694-35813694 on Build GRCh38
rs750288278 Chr.9: 35813791-35813791 on Build GRCh38
rs150427900 Chr.9: 35813762-35813762 on Build GRCh38
rs944351744 Chr.9: 35813787-35813787 on Build GRCh38
rs781729484 Chr.9: 35813703-35813703 on Build GRCh38
rs1014937763 Chr.9: 35813814-35813814 on Build GRCh38
rs985842085 Chr.9: 35813725-35813725 on Build GRCh38
rs780482955 Chr.9: 35813692-35813692 on Build GRCh38
rs755226170 Chr.9: 35813798-35813798 on Build GRCh38
rs554049277 Chr.9: 35813790-35813790 on Build GRCh38
rs370925110 Chr.9: 35813677-35813677 on Build GRCh38

View Full Product Details