Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68896380-68896625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL21
Entrez Gene ID
219927
Gene Name
mitochondrial ribosomal protein L21
Gene Aliases
L21mt, MRP-L21
UniGene
Hs.503047
Gene Chromosome Location
Chr.11: 68891276-68903835 on Build GRCh38
Transcripts
NM_181514.1 NM_181515.1 XM_005273823.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842830 c.386G>A p.R129Q Chr.11: 68896525-68896525 on Build GRCh38
5913224 c.329C>T p.S110F Chr.11: 68896582-68896582 on Build GRCh38
4878955 c.368C>T p.A123V Chr.11: 68896543-68896543 on Build GRCh38
6014535 c.396+8C>T p.? Chr.11: 68896507-68896507 on Build GRCh38
2043062 c.385C>T p.R129* Chr.11: 68896526-68896526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149807624 Chr.11: 68896434-68896434 on Build GRCh38
rs943783940 Chr.11: 68896451-68896451 on Build GRCh38
rs182603715 Chr.11: 68896467-68896467 on Build GRCh38
rs766488912 Chr.11: 68896530-68896531 on Build GRCh38
rs932523696 Chr.11: 68896480-68896480 on Build GRCh38
rs982457125 Chr.11: 68896433-68896433 on Build GRCh38
rs758539056 Chr.11: 68896605-68896605 on Build GRCh38
rs781353875 Chr.11: 68896516-68896516 on Build GRCh38
rs541136999 Chr.11: 68896508-68896508 on Build GRCh38
rs569529946 Chr.11: 68896454-68896454 on Build GRCh38
rs748442191 Chr.11: 68896439-68896439 on Build GRCh38
rs368358628 Chr.11: 68896503-68896503 on Build GRCh38
rs1041703239 Chr.11: 68896512-68896512 on Build GRCh38
rs146772580 Chr.11: 68896584-68896584 on Build GRCh38
rs979017784 Chr.11: 68896498-68896498 on Build GRCh38
rs758449147 Chr.11: 68896497-68896497 on Build GRCh38
rs578139438 Chr.11: 68896569-68896569 on Build GRCh38
rs553569438 Chr.11: 68896564-68896564 on Build GRCh38
rs867122505 Chr.11: 68896416-68896416 on Build GRCh38
rs190359911 Chr.11: 68896415-68896415 on Build GRCh38
rs372240224 Chr.11: 68896543-68896543 on Build GRCh38
rs148535951 Chr.11: 68896600-68896600 on Build GRCh38
rs564042523 Chr.11: 68896601-68896601 on Build GRCh38
rs143584803 Chr.11: 68896525-68896525 on Build GRCh38
rs976937734 Chr.11: 68896453-68896453 on Build GRCh38
rs764036906 Chr.11: 68896440-68896440 on Build GRCh38
rs968543867 Chr.11: 68896486-68896486 on Build GRCh38
rs768253705 Chr.11: 68896550-68896550 on Build GRCh38
rs747191783 Chr.11: 68896504-68896504 on Build GRCh38
rs1019700373 Chr.11: 68896421-68896421 on Build GRCh38
rs764066653 Chr.11: 68896607-68896607 on Build GRCh38
rs746382949 Chr.11: 68896523-68896523 on Build GRCh38
rs755975896 Chr.11: 68896466-68896466 on Build GRCh38
rs775164150 Chr.11: 68896542-68896542 on Build GRCh38
rs750383688 Chr.11: 68896464-68896464 on Build GRCh38
rs772838658 Chr.11: 68896574-68896574 on Build GRCh38
rs992841927 Chr.11: 68896526-68896526 on Build GRCh38
rs774123688 Chr.11: 68896557-68896557 on Build GRCh38
rs760625100 Chr.11: 68896599-68896599 on Build GRCh38
rs76789024 Chr.11: 68896484-68896484 on Build GRCh38
rs893730557 Chr.11: 68896494-68896494 on Build GRCh38

View Full Product Details