Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7497535-7497800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2A
Entrez Gene ID
5430
Gene Name
RNA polymerase II subunit A
Gene Aliases
POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
UniGene
Hs.270017
Gene Chromosome Location
Chr.17: 7484379-7514618 on Build GRCh38
Transcripts
NM_000937.4

SNP Detail

SNP ID Chromosome Location
rs148069571 Chr.17: 7497754-7497754 on Build GRCh38
rs771424087 Chr.17: 7497645-7497645 on Build GRCh38
rs750343151 Chr.17: 7497619-7497619 on Build GRCh38
rs759016984 Chr.17: 7497563-7497563 on Build GRCh38
rs1035681337 Chr.17: 7497588-7497588 on Build GRCh38
rs749734949 Chr.17: 7497642-7497642 on Build GRCh38
rs373929526 Chr.17: 7497769-7497769 on Build GRCh38
rs983604225 Chr.17: 7497576-7497576 on Build GRCh38
rs755035282 Chr.17: 7497623-7497623 on Build GRCh38
rs763858656 Chr.17: 7497601-7497601 on Build GRCh38
rs950982099 Chr.17: 7497573-7497573 on Build GRCh38
rs567340253 Chr.17: 7497721-7497721 on Build GRCh38
rs750332716 Chr.17: 7497730-7497730 on Build GRCh38
rs527303150 Chr.17: 7497685-7497685 on Build GRCh38
rs147691592 Chr.17: 7497691-7497691 on Build GRCh38
rs781161473 Chr.17: 7497625-7497625 on Build GRCh38
rs559854388 Chr.17: 7497667-7497667 on Build GRCh38
rs199782471 Chr.17: 7497567-7497567 on Build GRCh38
rs960962740 Chr.17: 7497598-7497598 on Build GRCh38
rs752936492 Chr.17: 7497628-7497628 on Build GRCh38
rs369235845 Chr.17: 7497647-7497647 on Build GRCh38
rs776271881 Chr.17: 7497692-7497692 on Build GRCh38
rs746396773 Chr.17: 7497650-7497650 on Build GRCh38
rs752877455 Chr.17: 7497775-7497775 on Build GRCh38
rs376608405 Chr.17: 7497604-7497604 on Build GRCh38
rs925693835 Chr.17: 7497641-7497641 on Build GRCh38
rs767612000 Chr.17: 7497764-7497764 on Build GRCh38
rs760363564 Chr.17: 7497599-7497599 on Build GRCh38
rs765157011 Chr.17: 7497607-7497607 on Build GRCh38
rs771889821 Chr.17: 7497609-7497610 on Build GRCh38
rs934494016 Chr.17: 7497683-7497683 on Build GRCh38
rs563906655 Chr.17: 7497673-7497673 on Build GRCh38
rs756450484 Chr.17: 7497630-7497630 on Build GRCh38
rs771828226 Chr.17: 7497562-7497563 on Build GRCh38
rs143227478 Chr.17: 7497713-7497713 on Build GRCh38
rs1013479223 Chr.17: 7497688-7497688 on Build GRCh38
rs978543859 Chr.17: 7497624-7497624 on Build GRCh38

View Full Product Details