Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 33186082-33186343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRP1
Entrez Gene ID
8829
Gene Name
neuropilin 1
Gene Aliases
BDCA4, CD304, NP1, NRP, VEGF165R
UniGene
Hs.131704
Gene Chromosome Location
Chr.10: 33177491-33334905 on Build GRCh38
Transcripts
NM_001244972.1 NM_001244973.1 NM_003873.5 NR_045259.1 XM_006717521.1 XM_006717522.1 XM_006717523.2 XM_006717524.1 XM_006717525.1 XM_017016865.1 XM_017016866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4013895 c.2233C>T p.P745S Chr.10: 33186318-33186318 on Build GRCh38
5029712 c.2300G>A p.R767H Chr.10: 33186251-33186251 on Build GRCh38
6729809 c.2273A>G p.H758R Chr.10: 33186278-33186278 on Build GRCh38
6282845 c.2256C>A p.V752V Chr.10: 33186295-33186295 on Build GRCh38
4013896 c.2222G>A p.R741H Chr.10: 33186329-33186329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961472873 Chr.10: 33186117-33186117 on Build GRCh38
rs781478517 Chr.10: 33186211-33186211 on Build GRCh38
rs1014030560 Chr.10: 33186283-33186283 on Build GRCh38
rs976861792 Chr.10: 33186121-33186121 on Build GRCh38
rs780027653 Chr.10: 33186289-33186289 on Build GRCh38
rs932706749 Chr.10: 33186131-33186131 on Build GRCh38
rs377600787 Chr.10: 33186189-33186189 on Build GRCh38
rs745403355 Chr.10: 33186276-33186276 on Build GRCh38
rs566477081 Chr.10: 33186306-33186306 on Build GRCh38
rs746985525 Chr.10: 33186297-33186297 on Build GRCh38
rs370641686 Chr.10: 33186272-33186272 on Build GRCh38
rs143147785 Chr.10: 33186110-33186110 on Build GRCh38
rs1027702130 Chr.10: 33186315-33186315 on Build GRCh38
rs761981034 Chr.10: 33186170-33186170 on Build GRCh38
rs755360383 Chr.10: 33186209-33186209 on Build GRCh38
rs768084911 Chr.10: 33186106-33186106 on Build GRCh38
rs879088621 Chr.10: 33186235-33186235 on Build GRCh38
rs1052901103 Chr.10: 33186213-33186213 on Build GRCh38
rs770032075 Chr.10: 33186307-33186307 on Build GRCh38
rs772069753 Chr.10: 33186284-33186284 on Build GRCh38
rs924149280 Chr.10: 33186252-33186252 on Build GRCh38
rs765276503 Chr.10: 33186184-33186184 on Build GRCh38
rs891653165 Chr.10: 33186239-33186239 on Build GRCh38
rs925900420 Chr.10: 33186159-33186159 on Build GRCh38
rs778519380 Chr.10: 33186273-33186273 on Build GRCh38
rs374109281 Chr.10: 33186186-33186186 on Build GRCh38
rs751816592 Chr.10: 33186202-33186202 on Build GRCh38
rs533049571 Chr.10: 33186114-33186114 on Build GRCh38
rs374865894 Chr.10: 33186302-33186302 on Build GRCh38
rs763405657 Chr.10: 33186187-33186187 on Build GRCh38
rs550737752 Chr.10: 33186142-33186142 on Build GRCh38
rs753501647 Chr.10: 33186230-33186230 on Build GRCh38
rs757021030 Chr.10: 33186237-33186237 on Build GRCh38
rs148799934 Chr.10: 33186251-33186251 on Build GRCh38
rs922669822 Chr.10: 33186122-33186122 on Build GRCh38

View Full Product Details