Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122172953-122173212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1109
Entrez Gene ID
84162
Gene Name
KIAA1109
Gene Aliases
FSA, Tweek
UniGene
Hs.408142
Gene Chromosome Location
Chr.4: 122152333-122362759 on Build GRCh38
Transcripts
NM_015312.3 XM_005263282.1 XM_005263287.1 XM_006714344.1 XM_011532319.1 XM_011532320.2 XM_011532321.2 XM_011532322.1 XM_011532323.1 XM_011532324.1 XM_011532325.1 XM_011532326.1 XM_011532327.1 XM_017008695.1 XM_017008696.1 XM_017008697.1 XM_017008699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5471811 c.98-4A>T p.? Chr.4: 122173032-122173032 on Build GRCh38
6693664 c.148C>T p.R50W Chr.4: 122173086-122173086 on Build GRCh38
4851375 c.211A>T p.I71F Chr.4: 122173149-122173149 on Build GRCh38
5908408 c.130C>T p.L44F Chr.4: 122173068-122173068 on Build GRCh38
5899461 c.146C>T p.S49F Chr.4: 122173084-122173084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768713955 Chr.4: 122173129-122173129 on Build GRCh38
rs763481775 Chr.4: 122173177-122173177 on Build GRCh38
rs971783075 Chr.4: 122173018-122173018 on Build GRCh38
rs750836903 Chr.4: 122172994-122172994 on Build GRCh38
rs745712120 Chr.4: 122173099-122173099 on Build GRCh38
rs762329807 Chr.4: 122173135-122173135 on Build GRCh38
rs752148128 Chr.4: 122173010-122173010 on Build GRCh38
rs951818166 Chr.4: 122173039-122173039 on Build GRCh38
rs1019839414 Chr.4: 122172992-122172992 on Build GRCh38
rs765654060 Chr.4: 122172990-122172990 on Build GRCh38
rs375832395 Chr.4: 122173008-122173008 on Build GRCh38
rs757174837 Chr.4: 122173063-122173063 on Build GRCh38
rs200536284 Chr.4: 122173081-122173081 on Build GRCh38
rs779122472 Chr.4: 122173049-122173050 on Build GRCh38
rs201902453 Chr.4: 122173019-122173019 on Build GRCh38
rs561937768 Chr.4: 122173114-122173114 on Build GRCh38
rs753744895 Chr.4: 122173023-122173023 on Build GRCh38
rs866512470 Chr.4: 122173048-122173049 on Build GRCh38
rs201127144 Chr.4: 122173005-122173005 on Build GRCh38
rs201312016 Chr.4: 122173124-122173124 on Build GRCh38
rs570830117 Chr.4: 122173017-122173017 on Build GRCh38
rs776603810 Chr.4: 122173130-122173130 on Build GRCh38
rs772276998 Chr.4: 122173107-122173107 on Build GRCh38
rs200085463 Chr.4: 122173159-122173159 on Build GRCh38
rs140628976 Chr.4: 122173160-122173160 on Build GRCh38

View Full Product Details