Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10446510-10446745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLRC1
Entrez Gene ID
3821
Gene Name
killer cell lectin like receptor C1
Gene Aliases
CD159A, NKG2, NKG2A
UniGene
Hs.512576
Gene Chromosome Location
Chr.12: 10442264-10454685 on Build GRCh38
Transcripts
NM_001304448.1 NM_002259.4 NM_007328.3 NM_213657.2 NM_213658.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6053388 c.696G>T p.K232N Chr.12: 10446557-10446557 on Build GRCh38
222236 c.682C>T p.H228Y Chr.12: 10446571-10446571 on Build GRCh38
6071120 c.653C>G p.S218* Chr.12: 10446600-10446600 on Build GRCh38
934324 c.702G>T p.*234Y Chr.12: 10446551-10446551 on Build GRCh38
3687916 c.629T>C p.V210A Chr.12: 10446624-10446624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768044829 Chr.12: 10446669-10446669 on Build GRCh38
rs372615172 Chr.12: 10446542-10446542 on Build GRCh38
rs144003798 Chr.12: 10446586-10446586 on Build GRCh38
rs752451278 Chr.12: 10446685-10446685 on Build GRCh38
rs746885638 Chr.12: 10446610-10446610 on Build GRCh38
rs775664960 Chr.12: 10446562-10446562 on Build GRCh38
rs778651698 Chr.12: 10446598-10446598 on Build GRCh38
rs139182795 Chr.12: 10446578-10446578 on Build GRCh38
rs747080236 Chr.12: 10446543-10446543 on Build GRCh38
rs753393506 Chr.12: 10446589-10446589 on Build GRCh38
rs762242349 Chr.12: 10446665-10446665 on Build GRCh38
rs771880793 Chr.12: 10446541-10446541 on Build GRCh38
rs776860612 Chr.12: 10446553-10446553 on Build GRCh38
rs781378713 Chr.12: 10446618-10446618 on Build GRCh38
rs7314384 Chr.12: 10446671-10446671 on Build GRCh38
rs764556321 Chr.12: 10446569-10446569 on Build GRCh38
rs754548593 Chr.12: 10446591-10446591 on Build GRCh38
rs777839485 Chr.12: 10446609-10446609 on Build GRCh38
rs752401508 Chr.12: 10446599-10446599 on Build GRCh38
rs771192910 Chr.12: 10446549-10446549 on Build GRCh38
rs1020612489 Chr.12: 10446626-10446626 on Build GRCh38
rs770992699 Chr.12: 10446617-10446617 on Build GRCh38
rs758136731 Chr.12: 10446693-10446693 on Build GRCh38
rs759125102 Chr.12: 10446680-10446680 on Build GRCh38
rs769071378 Chr.12: 10446645-10446645 on Build GRCh38
rs758332936 Chr.12: 10446607-10446607 on Build GRCh38
rs150596852 Chr.12: 10446561-10446561 on Build GRCh38
rs763226245 Chr.12: 10446644-10446644 on Build GRCh38
rs369864314 Chr.12: 10446698-10446698 on Build GRCh38
rs759606503 Chr.12: 10446556-10446556 on Build GRCh38
rs774712666 Chr.12: 10446664-10446664 on Build GRCh38
rs568051460 Chr.12: 10446620-10446620 on Build GRCh38
rs377273500 Chr.12: 10446571-10446571 on Build GRCh38
rs763426864 Chr.12: 10446568-10446568 on Build GRCh38
rs369788262 Chr.12: 10446684-10446684 on Build GRCh38
rs775825553 Chr.12: 10446631-10446631 on Build GRCh38

View Full Product Details