Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 36817703-36817928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNU1
Entrez Gene ID
157855
Gene Name
potassium calcium-activated channel subfamily U member 1
Gene Aliases
KCNMC1, KCa5, KCa5.1, Kcnma3, Slo3
UniGene
Hs.13861
Gene Chromosome Location
Chr.8: 36784324-36936125 on Build GRCh38
Transcripts
NM_001031836.2 NR_134267.1 XM_011544424.1 XM_011544426.1 XM_011544427.1 XM_017013144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373273 c.1072G>A p.G358R Chr.8: 36817726-36817726 on Build GRCh38
1719236 c.1059C>T p.L353L Chr.8: 36817713-36817713 on Build GRCh38
6892165 c.1050G>A p.R350R Chr.8: 36817704-36817704 on Build GRCh38
4883890 c.1070C>A p.S357* Chr.8: 36817724-36817724 on Build GRCh38
6897726 c.1099C>T p.L367L Chr.8: 36817753-36817753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767218544 Chr.8: 36817808-36817808 on Build GRCh38
rs772463602 Chr.8: 36817768-36817768 on Build GRCh38
rs749809724 Chr.8: 36817735-36817735 on Build GRCh38
rs756462308 Chr.8: 36817724-36817724 on Build GRCh38
rs752363863 Chr.8: 36817809-36817809 on Build GRCh38
rs529652011 Chr.8: 36817779-36817779 on Build GRCh38
rs371231739 Chr.8: 36817810-36817810 on Build GRCh38
rs754438364 Chr.8: 36817800-36817800 on Build GRCh38
rs551303900 Chr.8: 36817785-36817785 on Build GRCh38
rs745914696 Chr.8: 36817744-36817744 on Build GRCh38
rs751116457 Chr.8: 36817791-36817791 on Build GRCh38
rs947740749 Chr.8: 36817813-36817813 on Build GRCh38
rs760922317 Chr.8: 36817776-36817776 on Build GRCh38
rs980887202 Chr.8: 36817840-36817840 on Build GRCh38
rs752698221 Chr.8: 36817888-36817888 on Build GRCh38
rs778056016 Chr.8: 36817727-36817727 on Build GRCh38
rs773100958 Chr.8: 36817782-36817782 on Build GRCh38
rs775841062 Chr.8: 36817770-36817770 on Build GRCh38
rs771304434 Chr.8: 36817736-36817736 on Build GRCh38
rs748583177 Chr.8: 36817723-36817723 on Build GRCh38
rs1047606428 Chr.8: 36817899-36817899 on Build GRCh38
rs1052191901 Chr.8: 36817897-36817897 on Build GRCh38
rs911293703 Chr.8: 36817764-36817764 on Build GRCh38
rs774513283 Chr.8: 36817742-36817742 on Build GRCh38
rs1038370414 Chr.8: 36817869-36817869 on Build GRCh38
rs993080940 Chr.8: 36817757-36817757 on Build GRCh38
rs540045455 Chr.8: 36817872-36817872 on Build GRCh38
rs1050367662 Chr.8: 36817732-36817732 on Build GRCh38
rs557533545 Chr.8: 36817895-36817895 on Build GRCh38

View Full Product Details