Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65052933-65053204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASL12
Entrez Gene ID
51285
Gene Name
RAS like family 12
Gene Aliases
RIS
UniGene
Hs.27018
Gene Chromosome Location
Chr.15: 65045576-65076654 on Build GRCh38
Transcripts
XM_005254434.4 XM_017022296.1 NM_178859.3 XM_005254159.4

Gene Symbol
SLC51B
Entrez Gene ID
123264
Gene Name
solute carrier family 51 beta subunit
Gene Aliases
OSTB, OSTBETA
UniGene
Hs.27018
Gene Chromosome Location
Chr.15: 65045370-65053396 on Build GRCh38
Transcripts
XM_005254434.4 XM_017022296.1 NM_178859.3 XM_005254159.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000475 c.205C>A p.P69T Chr.15: 65052982-65052982 on Build GRCh38
4710479 c.200T>C p.M67T Chr.15: 65052977-65052977 on Build GRCh38
963996 c.193G>T p.E65* Chr.15: 65052970-65052970 on Build GRCh38
7390704 c.314A>C p.Q105P Chr.15: 65053091-65053091 on Build GRCh38
4541211 c.295G>T p.E99* Chr.15: 65053072-65053072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768918597 Chr.15: 65053044-65053045 on Build GRCh38
rs747393047 Chr.15: 65053083-65053083 on Build GRCh38
rs969652666 Chr.15: 65053168-65053168 on Build GRCh38
rs773438957 Chr.15: 65053125-65053125 on Build GRCh38
rs768878212 Chr.15: 65053098-65053098 on Build GRCh38
rs377515714 Chr.15: 65052960-65052960 on Build GRCh38
rs150408669 Chr.15: 65052976-65052976 on Build GRCh38
rs751403585 Chr.15: 65053035-65053035 on Build GRCh38
rs747097963 Chr.15: 65052964-65052968 on Build GRCh38
rs780226546 Chr.15: 65053044-65053044 on Build GRCh38
rs776892359 Chr.15: 65053053-65053053 on Build GRCh38
rs773595808 Chr.15: 65053110-65053111 on Build GRCh38
rs765197466 Chr.15: 65052998-65052998 on Build GRCh38
rs748597979 Chr.15: 65053111-65053111 on Build GRCh38
rs777264075 Chr.15: 65053063-65053063 on Build GRCh38
rs865956343 Chr.15: 65053169-65053169 on Build GRCh38
rs201580924 Chr.15: 65053002-65053002 on Build GRCh38
rs994236086 Chr.15: 65052961-65052961 on Build GRCh38
rs574380553 Chr.15: 65053001-65053001 on Build GRCh38
rs763965192 Chr.15: 65053175-65053175 on Build GRCh38
rs772899639 Chr.15: 65053140-65053140 on Build GRCh38
rs752749624 Chr.15: 65053158-65053158 on Build GRCh38
rs770588345 Chr.15: 65053068-65053068 on Build GRCh38
rs1054962927 Chr.15: 65053064-65053064 on Build GRCh38
rs533867122 Chr.15: 65053017-65053017 on Build GRCh38
rs748379131 Chr.15: 65053062-65053063 on Build GRCh38
rs937862730 Chr.15: 65053055-65053055 on Build GRCh38
rs749002494 Chr.15: 65053067-65053067 on Build GRCh38
rs778493381 Chr.15: 65053069-65053069 on Build GRCh38
rs767187250 Chr.15: 65053146-65053146 on Build GRCh38
rs755993629 Chr.15: 65053170-65053170 on Build GRCh38
rs960821237 Chr.15: 65053023-65053023 on Build GRCh38
rs752556839 Chr.15: 65053044-65053044 on Build GRCh38
rs759426278 Chr.15: 65053144-65053144 on Build GRCh38
rs142436732 Chr.15: 65053106-65053106 on Build GRCh38
rs913552152 Chr.15: 65052969-65052969 on Build GRCh38
rs80035684 Chr.15: 65052958-65052958 on Build GRCh38
rs770073689 Chr.15: 65053108-65053111 on Build GRCh38
rs776952059 Chr.15: 65052963-65052963 on Build GRCh38
rs147682812 Chr.15: 65053049-65053049 on Build GRCh38
rs75962063 Chr.15: 65053139-65053139 on Build GRCh38
rs763251825 Chr.15: 65053018-65053018 on Build GRCh38
rs781214644 Chr.15: 65053038-65053038 on Build GRCh38
rs766649904 Chr.15: 65053024-65053024 on Build GRCh38

View Full Product Details