Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115286410-115286665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGF
Entrez Gene ID
4803
Gene Name
nerve growth factor
Gene Aliases
Beta-NGF, HSAN5, NGFB
UniGene
Hs.2561
Gene Chromosome Location
Chr.1: 115285915-115338253 on Build GRCh38
Transcripts
NM_002506.2 XM_006710663.3 XM_011541518.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5901062 c.376C>T p.P126S Chr.1: 115286420-115286420 on Build GRCh38
2177896 c.155G>A p.R52H Chr.1: 115286641-115286641 on Build GRCh38
4170535 c.281C>T p.P94L Chr.1: 115286515-115286515 on Build GRCh38
2177893 c.170C>T p.A57V Chr.1: 115286626-115286626 on Build GRCh38
221486 c.160G>A p.A54T Chr.1: 115286636-115286636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750030065 Chr.1: 115286478-115286478 on Build GRCh38
rs147326889 Chr.1: 115286622-115286622 on Build GRCh38
rs372994894 Chr.1: 115286589-115286589 on Build GRCh38
rs766912679 Chr.1: 115286642-115286642 on Build GRCh38
rs142538674 Chr.1: 115286497-115286497 on Build GRCh38
rs377253859 Chr.1: 115286593-115286593 on Build GRCh38
rs767878560 Chr.1: 115286608-115286608 on Build GRCh38
rs767272925 Chr.1: 115286548-115286548 on Build GRCh38
rs766082334 Chr.1: 115286600-115286600 on Build GRCh38
rs766849758 Chr.1: 115286553-115286553 on Build GRCh38
rs572066909 Chr.1: 115286594-115286594 on Build GRCh38
rs750836533 Chr.1: 115286640-115286640 on Build GRCh38
rs753382007 Chr.1: 115286555-115286555 on Build GRCh38
rs372694067 Chr.1: 115286487-115286487 on Build GRCh38
rs748148537 Chr.1: 115286620-115286620 on Build GRCh38
rs759051358 Chr.1: 115286604-115286604 on Build GRCh38
rs267597942 Chr.1: 115286545-115286545 on Build GRCh38
rs200459956 Chr.1: 115286636-115286636 on Build GRCh38
rs761709969 Chr.1: 115286544-115286544 on Build GRCh38
rs768548505 Chr.1: 115286630-115286636 on Build GRCh38
rs758948909 Chr.1: 115286498-115286498 on Build GRCh38
rs775429651 Chr.1: 115286454-115286454 on Build GRCh38
rs201861727 Chr.1: 115286623-115286623 on Build GRCh38
rs138175552 Chr.1: 115286549-115286549 on Build GRCh38
rs757830779 Chr.1: 115286607-115286607 on Build GRCh38
rs376241395 Chr.1: 115286637-115286637 on Build GRCh38
rs755243469 Chr.1: 115286612-115286612 on Build GRCh38
rs760350868 Chr.1: 115286595-115286595 on Build GRCh38
rs774379026 Chr.1: 115286540-115286540 on Build GRCh38
rs751440257 Chr.1: 115286464-115286464 on Build GRCh38
rs759547617 Chr.1: 115286433-115286433 on Build GRCh38
rs968846429 Chr.1: 115286554-115286554 on Build GRCh38
rs145894043 Chr.1: 115286474-115286474 on Build GRCh38
rs776406021 Chr.1: 115286628-115286628 on Build GRCh38
rs770647680 Chr.1: 115286626-115286626 on Build GRCh38
rs778266438 Chr.1: 115286558-115286558 on Build GRCh38
rs537372354 Chr.1: 115286504-115286504 on Build GRCh38
rs771270154 Chr.1: 115286513-115286513 on Build GRCh38
rs149054069 Chr.1: 115286641-115286641 on Build GRCh38
rs867581456 Chr.1: 115286456-115286456 on Build GRCh38
rs202005277 Chr.1: 115286475-115286475 on Build GRCh38
rs972935103 Chr.1: 115286565-115286565 on Build GRCh38
rs751997242 Chr.1: 115286567-115286567 on Build GRCh38
rs780432648 Chr.1: 115286610-115286610 on Build GRCh38
rs1035937750 Chr.1: 115286489-115286489 on Build GRCh38
rs776208737 Chr.1: 115286603-115286603 on Build GRCh38
rs746169118 Chr.1: 115286531-115286531 on Build GRCh38
rs11466111 Chr.1: 115286557-115286557 on Build GRCh38
rs775686730 Chr.1: 115286534-115286534 on Build GRCh38
rs150136942 Chr.1: 115286512-115286512 on Build GRCh38
rs1043758056 Chr.1: 115286434-115286434 on Build GRCh38
rs755580704 Chr.1: 115286480-115286480 on Build GRCh38
rs369266810 Chr.1: 115286611-115286611 on Build GRCh38
rs35941329 Chr.1: 115286460-115286460 on Build GRCh38
rs779678303 Chr.1: 115286481-115286481 on Build GRCh38
rs149876217 Chr.1: 115286631-115286631 on Build GRCh38
rs11466110 Chr.1: 115286582-115286582 on Build GRCh38
rs746837424 Chr.1: 115286625-115286625 on Build GRCh38
rs200629339 Chr.1: 115286539-115286539 on Build GRCh38
rs765918245 Chr.1: 115286550-115286550 on Build GRCh38
rs202060337 Chr.1: 115286520-115286520 on Build GRCh38
rs147763877 Chr.1: 115286461-115286461 on Build GRCh38
rs141486298 Chr.1: 115286572-115286572 on Build GRCh38
rs757701792 Chr.1: 115286571-115286571 on Build GRCh38
rs779189568 Chr.1: 115286616-115286616 on Build GRCh38
rs775019570 Chr.1: 115286632-115286632 on Build GRCh38
rs769893063 Chr.1: 115286532-115286532 on Build GRCh38
rs780383333 Chr.1: 115286577-115286577 on Build GRCh38
rs750829893 Chr.1: 115286609-115286609 on Build GRCh38
rs754238855 Chr.1: 115286646-115286646 on Build GRCh38
rs756396299 Chr.1: 115286574-115286574 on Build GRCh38
rs768794852 Chr.1: 115286588-115286588 on Build GRCh38
rs201087374 Chr.1: 115286605-115286605 on Build GRCh38
rs376618971 Chr.1: 115286453-115286453 on Build GRCh38

View Full Product Details