Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197826651-197826918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH3
Entrez Gene ID
84859
Gene Name
leucine rich repeats and calponin homology domain containing 3
Gene Aliases
-
UniGene
Hs.518414
Gene Chromosome Location
Chr.3: 197790855-197889346 on Build GRCh38
Transcripts
NM_032773.3 XM_005269362.2 XM_005269363.2 XM_005269365.2 XM_005269367.2 XM_006713791.3 XM_006713792.3 XM_006713793.2 XM_011513242.2 XM_011513243.2 XM_017007350.1 XM_017007351.1 XM_017007352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6325088 c.666G>A p.R222R Chr.3: 197826903-197826903 on Build GRCh38
1617424 c.650A>G p.E217G Chr.3: 197826887-197826887 on Build GRCh38
6476605 c.664C>T p.R222W Chr.3: 197826901-197826901 on Build GRCh38
1566836 c.643C>T p.L215L Chr.3: 197826880-197826880 on Build GRCh38
5080493 c.643C>T p.L215L Chr.3: 197826880-197826880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770707908 Chr.3: 197826839-197826839 on Build GRCh38
rs893141184 Chr.3: 197826787-197826787 on Build GRCh38
rs371394845 Chr.3: 197826887-197826887 on Build GRCh38
rs761793969 Chr.3: 197826881-197826881 on Build GRCh38
rs1050907742 Chr.3: 197826769-197826769 on Build GRCh38
rs932079426 Chr.3: 197826757-197826757 on Build GRCh38
rs957664083 Chr.3: 197826683-197826683 on Build GRCh38
rs150116575 Chr.3: 197826884-197826884 on Build GRCh38
rs535174473 Chr.3: 197826686-197826686 on Build GRCh38
rs776313931 Chr.3: 197826781-197826781 on Build GRCh38
rs377726765 Chr.3: 197826885-197826885 on Build GRCh38
rs768214986 Chr.3: 197826711-197826711 on Build GRCh38
rs745784665 Chr.3: 197826860-197826860 on Build GRCh38
rs943884956 Chr.3: 197826710-197826710 on Build GRCh38
rs778601922 Chr.3: 197826851-197826851 on Build GRCh38
rs772184233 Chr.3: 197826877-197826877 on Build GRCh38
rs746656278 Chr.3: 197826684-197826684 on Build GRCh38
rs775125432 Chr.3: 197826879-197826879 on Build GRCh38
rs923380090 Chr.3: 197826732-197826732 on Build GRCh38
rs929361682 Chr.3: 197826746-197826756 on Build GRCh38
rs908440721 Chr.3: 197826695-197826695 on Build GRCh38
rs879004386 Chr.3: 197826731-197826738 on Build GRCh38
rs749482099 Chr.3: 197826828-197826828 on Build GRCh38
rs779116915 Chr.3: 197826865-197826866 on Build GRCh38
rs903423736 Chr.3: 197826845-197826845 on Build GRCh38

View Full Product Details