Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152350714-152350979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Gene Symbol
FLG2
Entrez Gene ID
388698
Gene Name
filaggrin family member 2
Gene Aliases
IFPS
UniGene
Hs.156124
Gene Chromosome Location
Chr.1: 152348735-152360028 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5796384 c.6878G>C p.R2293T Chr.1: 152350908-152350908 on Build GRCh38
3801896 c.6917G>C p.R2306T Chr.1: 152350869-152350869 on Build GRCh38
6540882 c.6876G>A p.G2292G Chr.1: 152350910-152350910 on Build GRCh38
391818 c.6923_6926delGCCA p.G2308fs*>83 Chr.1: 152350860-152350863 on Build GRCh38
3474094 c.7005G>A p.R2335R Chr.1: 152350781-152350781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117998963 Chr.1: 152350745-152350745 on Build GRCh38
rs201036750 Chr.1: 152350848-152350848 on Build GRCh38
rs909770614 Chr.1: 152350807-152350807 on Build GRCh38
rs767573635 Chr.1: 152350924-152350924 on Build GRCh38
rs765472682 Chr.1: 152350740-152350740 on Build GRCh38
rs746645123 Chr.1: 152350832-152350832 on Build GRCh38
rs757517293 Chr.1: 152350931-152350934 on Build GRCh38
rs369042064 Chr.1: 152350793-152350793 on Build GRCh38
rs750341551 Chr.1: 152350945-152350945 on Build GRCh38
rs773806773 Chr.1: 152350881-152350881 on Build GRCh38
rs751792163 Chr.1: 152350901-152350901 on Build GRCh38
rs940728888 Chr.1: 152350903-152350903 on Build GRCh38
rs766450777 Chr.1: 152350899-152350899 on Build GRCh38
rs762378859 Chr.1: 152350739-152350739 on Build GRCh38
rs768970100 Chr.1: 152350836-152350836 on Build GRCh38
rs376710514 Chr.1: 152350769-152350769 on Build GRCh38
rs140628166 Chr.1: 152350822-152350822 on Build GRCh38
rs766729553 Chr.1: 152350762-152350762 on Build GRCh38
rs567121398 Chr.1: 152350846-152350846 on Build GRCh38
rs977754979 Chr.1: 152350764-152350764 on Build GRCh38
rs149170140 Chr.1: 152350919-152350919 on Build GRCh38
rs1051057654 Chr.1: 152350953-152350953 on Build GRCh38
rs775057083 Chr.1: 152350829-152350829 on Build GRCh38
rs754206719 Chr.1: 152350751-152350761 on Build GRCh38
rs138450657 Chr.1: 152350951-152350951 on Build GRCh38
rs996267932 Chr.1: 152350906-152350906 on Build GRCh38
rs532713977 Chr.1: 152350877-152350877 on Build GRCh38
rs762212548 Chr.1: 152350868-152350868 on Build GRCh38
rs984939450 Chr.1: 152350865-152350865 on Build GRCh38
rs757195711 Chr.1: 152350806-152350806 on Build GRCh38
rs753667523 Chr.1: 152350926-152350926 on Build GRCh38
rs1036382877 Chr.1: 152350904-152350904 on Build GRCh38
rs777046165 Chr.1: 152350736-152350736 on Build GRCh38
rs941285145 Chr.1: 152350809-152350809 on Build GRCh38
rs150260227 Chr.1: 152350957-152350957 on Build GRCh38
rs758512952 Chr.1: 152350755-152350755 on Build GRCh38
rs142283343 Chr.1: 152350815-152350815 on Build GRCh38
rs763172922 Chr.1: 152350882-152350882 on Build GRCh38
rs757009784 Chr.1: 152350933-152350933 on Build GRCh38
rs948280692 Chr.1: 152350855-152350855 on Build GRCh38
rs201988010 Chr.1: 152350909-152350909 on Build GRCh38
rs778848779 Chr.1: 152350935-152350935 on Build GRCh38
rs755900803 Chr.1: 152350791-152350791 on Build GRCh38
rs183397787 Chr.1: 152350812-152350812 on Build GRCh38

View Full Product Details