Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28735151-28735424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK8
Entrez Gene ID
284086
Gene Name
NIMA related kinase 8
Gene Aliases
JCK, NEK12A, NPHP9, RHPD2
UniGene
Hs.448468
Gene Chromosome Location
Chr.17: 28727899-28743457 on Build GRCh38
Transcripts
NM_178170.2 XM_011524638.2 XM_011524640.2 XM_017024499.1 XM_017024500.1 XM_017024501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6080761 c.616G>A p.A206T Chr.17: 28735369-28735369 on Build GRCh38
7172079 c.487-1G>A p.? Chr.17: 28735239-28735239 on Build GRCh38
7261637 c.525T>C p.C175C Chr.17: 28735278-28735278 on Build GRCh38
4634177 c.565C>A p.L189M Chr.17: 28735318-28735318 on Build GRCh38
7078643 c.610G>A p.E204K Chr.17: 28735363-28735363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756486155 Chr.17: 28735332-28735332 on Build GRCh38
rs917763885 Chr.17: 28735179-28735179 on Build GRCh38
rs778770826 Chr.17: 28735371-28735371 on Build GRCh38
rs928092875 Chr.17: 28735191-28735191 on Build GRCh38
rs867749493 Chr.17: 28735174-28735174 on Build GRCh38
rs1056787465 Chr.17: 28735283-28735283 on Build GRCh38
rs878915879 Chr.17: 28735311-28735311 on Build GRCh38
rs746862817 Chr.17: 28735192-28735192 on Build GRCh38
rs370732749 Chr.17: 28735231-28735231 on Build GRCh38
rs11649957 Chr.17: 28735383-28735383 on Build GRCh38
rs752331372 Chr.17: 28735336-28735336 on Build GRCh38
rs775602574 Chr.17: 28735203-28735204 on Build GRCh38
rs758136571 Chr.17: 28735339-28735339 on Build GRCh38
rs774429112 Chr.17: 28735260-28735260 on Build GRCh38
rs550716433 Chr.17: 28735385-28735385 on Build GRCh38
rs760667292 Chr.17: 28735204-28735205 on Build GRCh38
rs764097839 Chr.17: 28735241-28735241 on Build GRCh38
rs201773965 Chr.17: 28735335-28735335 on Build GRCh38
rs555501870 Chr.17: 28735362-28735362 on Build GRCh38
rs768102953 Chr.17: 28735296-28735296 on Build GRCh38
rs750935779 Chr.17: 28735331-28735331 on Build GRCh38
rs915615007 Chr.17: 28735298-28735299 on Build GRCh38
rs758210798 Chr.17: 28735401-28735401 on Build GRCh38
rs762177713 Chr.17: 28735273-28735273 on Build GRCh38
rs938474085 Chr.17: 28735229-28735229 on Build GRCh38
rs769628597 Chr.17: 28735227-28735227 on Build GRCh38
rs756231248 Chr.17: 28735342-28735342 on Build GRCh38
rs756755633 Chr.17: 28735363-28735363 on Build GRCh38
rs777365994 Chr.17: 28735350-28735350 on Build GRCh38
rs770920490 Chr.17: 28735198-28735198 on Build GRCh38
rs978302716 Chr.17: 28735182-28735182 on Build GRCh38
rs776247740 Chr.17: 28735221-28735221 on Build GRCh38
rs745641602 Chr.17: 28735223-28735223 on Build GRCh38

View Full Product Details